Skip to Content
Merck
All Photos(7)

Documents

1.04374

Supelco

Hexane

≥98.50% (hexane isomers + methylcyclopentane, GC), ≥96.0% (GC), ACS reagent, reag. Ph. Eur., EMSURE®

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-777-6
NACRES:
NB.21

product name

n-Hexane, for analysis EMSURE® ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

product line

EMSURE®

Assay

≥96.0% (GC)
≥98.50% (hexane isomers + methylcyclopentane, GC)

form

liquid

autoignition temp.

453 °F

potency

25000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.7 %

impurities

≤0.0003 meq/g Water-soluble titrable acid
≤0.005% Sulfur compounds (as S)
≤0.01% Aromatics (as benzene)
≤0.01% Water

evapn. residue

≤0.001%

color

APHA: ≤10

transmittance

260-420 nm, ≥97%

refractive index

n20/D 1.375 (lit.)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

density

0.659 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

SMILES string

CCCCCC

storage temp.

2-30°C

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Oil produced from Ghana Shea Nut crop for prospective industrial applications.: This study explores the industrial applications of oil derived from Ghana′s Shea Nut crop, using n-hexane as a solvent for oil extraction. It discusses the properties of the oil, potential uses, and the role of n-hexane in obtaining high purity extracts (Alale et al., 2024).

  • Bio-guided isolation of a new sesquiterpene from Artemisia cina with anthelmintic activity against Haemonchus contortus L3 infective larvae.: This research highlights the use of n-hexane in the extraction and isolation of bioactive compounds, specifically a new sesquiterpene with potential anthelmintic properties, demonstrating n-hexane′s effectiveness in phytochemical extraction (Arango-De la Pava et al., 2024).

  • Rapid purification and scale-up of tilianin using counter-current chromatography with rectangular horizontal tubing.: Discusses the use of n-hexane in counter-current chromatography for the rapid purification and scale-up of tilianin, a bioactive compound, showcasing the solvent′s role in enhancing the efficiency of chromatographic processes (Zhang et al., 2024).

  • Phytochemicals, Antioxidant, and Antidiabetic Effects of Ranunculus hirtellus Aerial Parts and Roots: Methanol and Aqueous Extracts.: While primarily focusing on methanol and aqueous extracts, this study also discusses the comparative effectiveness of different solvents including n-hexane for extracting bioactive compounds from Ranunculus hirtellus, highlighting its potential in pharmacological studies (Ishtiaq et al., 2024).


Analysis Note

Purity (GC): ≥ 96.0 %
Purity Σ hexane isomers + methylcyclopentane (GC): ≥ 98.5 %
Identity (IR): conforms
Color: ≤ 10 Hazen
Water-soluble titrable acid: ≤ 0.0003 meq/g
Refractive index (n 20/D): 1.375 - 1.376
Density (d 20 °C/20 °C): 0.659 - 0.663
Boiling range (min.95 %): 67 - 69 °C
Transmission (between 260 nm and 420 nm): ≥ 97 %
Thiophene: conforms
Aromatics (as benzene): ≤ 0.01 %
Sulfur compounds (as S): ≤ 0.005 %
Readily carbonizable substances: conforms
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.01 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service