Skip to Content
Merck
All Photos(1)

Key Documents

1.04184

Sigma-Aldrich

Cresyl violet (acetate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N3O3
CAS Number:
Molecular Weight:
321.33
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

composition

Dye, ≥70% spectrophotometric assay

Quality Level

pH

3 (20 °C, 10 g/L in H2O)

mp

140-143 °C

solubility

70 g/L

bulk density

460 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

Nc1c2c(c3nc4c([o+]c3c1)cc(cc4)N)cccc2.[O-]C(=O)C

InChI

1S/C16H12N3O.C2H4O2/c17-9-5-6-13-14(7-9)20-15-8-12(18)10-3-1-2-4-11(10)16(15)19-13;1-2(3)4/h1-8H,17-18H2;1H3,(H,3,4)/q+1;/p-1

InChI key

XKOCOMKJPWEOHX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye Cresyl violet (acetate) for microscopy Certistain®, is used for the preparation of a staining solution that can be used for the staining of nuclei and Nissl bodies. Cresyl violet (acetate) belongs to the group of oxazine dyes. As a CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (spectrophotometrically): ≥ 70%
Identity
- acetate:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (ethanol 50%): 596 - 599 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; ethanol 50%): 980 - 1400
TLC-Test: passes test
Loss on drying (50 °C / <1 hPa): ≤ 25%
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service