Skip to Content
Merck
All Photos(4)

Key Documents

1.03115

Supelco

1,4-Dioxane

EMPLURA®

Synonym(s):

1,4-Dioxane, Glycolethylether, 1,4-Diethylene dioxide, 1,4-Dioxacyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12191504
EC Index Number:
204-661-8
NACRES:
NA.21
bp:
101.5 °C/1013 hPa
vapor pressure:
41 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

41 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

300 °C

potency

5200 mg/kg LD50, oral (Rat)
7600 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-25.2 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.005% Peroxide (as H2O2)
≤0.01% Free acid (as CH3COOH)
≤0.1% Water

pH

6-8 (20 °C, 500 g/L in H2O)

bp

101.5 °C/1013 hPa

mp

12 °C

transition temp

flash point 11 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1CCOCC1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Occurrence of 1,4-dioxane in drinking water of China and its ecological and human health risk: An examination of the prevalence of 1,4-dioxane in China′s drinking water, with an assessment of the associated ecological and human health risks, relevant for researchers and policymakers focused on public health and environmental safety (Wang et al., 2024).

  • Adsorption Behavior and Kinetics of 1,4-Dioxane by Carbon Aerogel: This research discusses the effectiveness of carbon aerogel in the adsorption of 1,4-dioxane, providing valuable data on adsorption kinetics and behavior, useful for material scientists and chemists working with advanced materials (Lu et al., 2024).

  • Photosensitized covalent organic framework as a light-induced oxidase mimic for colorimetric detection of uric acid: This innovative approach using a framework that could potentially be adapted for 1,4-dioxane detection underscores the intersection of material science and analytical chemistry, which could intrigue researchers developing new analytical methods (Wang et al., 2024).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.01 %
Density (d 20 °C/ 4 °C): 1.032 - 1.034
Peroxide (as H₂O₂): ≤ 0.005 %
Water: ≤ 0.1 %
Stabilized with about 25 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT)

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service