1.02023
LICHROLUT® RP-18 solid phase extraction (SPE) tubes
(40 - 63 µm) 500mg 3 ml, package of 50 tubes
Synonym(s):
RP-18 SPE Tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115711
NACRES:
NB.21
Recommended Products
Product Name
LiChrolut® RP-18 (40 - 63 µm) 500 mg 3 ml standard PP-tubes, package of 50 tubes
product line
LiChrolut®
packaging
package of 50 tubes
technique(s)
solid phase extraction (SPE): suitable
storage temp.
no temp limit
General description
50 extraction tubes per package
Lichrolut columns are designed for reliable and rapid solid phase extraction. This is achieved by using strong but reversible interactions between the analyte and the surface of the stationary phase. A variety of surface modifications allow non-polar extractions (Lichrolut RP18 and RP18e), polar extractions (Lichrolut Si and Lichrolut CN), Cation Exchange extraction (Lichrolut SCX), mixed mode extraction (Lichrolut TSC) and non-polar extractions on a polymer phase (Lichrolut EN) which is especially well suited for the extraction of pesticdes and phenols in water or drugs in body fluids.
Analysis Note
Appearance: conforms
Legal Information
LICHROLUT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service