1.01005
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF
Pharma Manufacturing
Synonym(s):
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, Acetone chloroform, Chlorbutol, Chloretone, Chlorobutanol
About This Item
Recommended Products
Agency
BP
JP
NF
Ph. Eur.
Quality Level
product line
EMPROVE® ESSENTIAL
form
solid
pH
4.5-6 (20 °C, 7.7 g/L in H2O)
bp
167 °C/1013 hPa
mp
78 °C
transition temp
flash point 100 °C
solubility
7.7 g/L
application(s)
liquid formulation
ophthalmics
pharmaceutical
storage temp.
15-25°C
SMILES string
ClC(Cl)(Cl)C(O)(C)C.ClC(Cl)(Cl)C(O)(C)C.O
InChI
1S/2C4H7Cl3O.H2O/c2*1-3(2,8)4(5,6)7;/h2*8H,1-2H3;1H2
InChI key
WRWLCXJYIMRJIN-UHFFFAOYSA-N
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
Disclaimer
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
212.0 °F - closed cup
Flash Point(C)
100 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service