Skip to Content
Merck

1.00635

SAFC

Salicylic acid

fine powder EMPROVE® ESSENTIAL Ph Eur,BP,USP

Pharma Manufacturing

Synonym(s):

Salicylic acid, 2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-712-3
Pricing and availability is not currently available.

Agency

BP
Ph. Eur.
USP

Quality Level

vapor pressure

<1 hPa ( 100 °C)

product line

EMPROVE® ESSENTIAL

Assay

98.0-102.0% dry basis (HPLC)

form

solid

autoignition temp.

500 °C

potency

1250-1580 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

technique(s)

tissue culture: suitable

pH

2.4 ( in H2O, saturated solution)

bp

211 °C/1013 hPa

mp

157-159 °C

transition temp

flash point 157 °C

solubility

water: 2 g/L at 20 °C

density

1.443 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

anion traces

chloride (Cl-): ≤0.0100%
sulfate (SO42-): ≤0.0200%

application(s)

liquid formulation
pharmaceutical

storage temp.

15-25°C

SMILES string

[O-]c1c(cccc1)C(=O)O.[H+]

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Salicylic acid is used in dermatological medicine for skin exfoliating and acne. Salicylic acid Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service