Skip to Content
Merck
All Photos(3)

Documents

1.00316

Supelco

Hydrochloric acid

25%, for analysis EMSURE®

Synonym(s):

Hydrochloric acid solution, Hydrochloric acid 25%, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

product name

Hydrochloric acid solution, 25% for analysis EMSURE®

vapor pressure

12 hPa ( 20 °C)

Quality Level

product line

EMSURE®

Assay

≥25.0% (alkalimetric)

form

liquid

impurities

≤0.01 ppm As (Arsenic)
≤0.010 ppm Cd (Cadmium)
≤0.010 ppm Co (Cobalt)
≤0.010 ppm Li (Lithium)
≤0.010 ppm Mn (Manganese)
≤0.010 ppm Sr (Strontium)
≤0.010 ppm V (Vanadium)
≤0.020 ppm Ba (Barium)
≤0.020 ppm Be (Beryllium)
≤0.020 ppm Cr (Chromium)
≤0.020 ppm Cu (Copper)
≤0.020 ppm Mo (Molybdenum)
≤0.020 ppm Ni (Nickel)
≤0.020 ppm Pb (Lead)
≤0.050 ppm Al (Aluminium)
≤0.050 ppm Ge (Germanium)
≤0.050 ppm Tl (Thallium)
≤0.050 ppm Zn (Zinc)
≤0.100 ppm Bi (Bismuth)
≤0.100 ppm K (Potassium)
≤0.100 ppm Mg (Magnesium)
≤0.100 ppm Ti (Titanium)
≤0.100 ppm Zr (Zirconium)
≤0.200 ppm Fe (Iron)
≤0.5 ppm Free chlorine (Cl)
≤0.500 ppm Ca (Calcium)
≤0.500 ppm Na (Sodium)
≤1 ppm NH4 (Ammonium)
≤1 ppm Sulfate (SO4)
≤1 ppm Sulfite (SO3)
≤50 ppm Bromide (Br)

ign. residue

≤5 ppm (as SO4)

color

APHA: ≤10

pH

<1 (20 °C in H2O)

kinematic viscosity

1.73 cSt(20 °C)

bp

107 °C/1013 hPa

SMILES string

Cl

mp

-52 °C

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

for analysis EMSURE®

Application


  • Research on the Synthesis of Zinc-Ammonium Phosphate Using Galvanic Waste Sludge as a Source of Zinc: This study explores a novel approach using hydrochloric acid solution in the synthesis process aimed at valorizing waste materials. The article demonstrates a chemical strategy that enhances resource recovery, significantly contributing to waste minimization in industrial practices (Morgovan et al., 2024).

  • Contactless conductivity sensor employing moist paper as absorbent for in-situ detection of generated carbon dioxide gas: The study integrates hydrochloric acid solutions in the development of a novel, cost-effective contactless conductivity sensor. This sensor uses a simple paper-based system for the in-situ monitoring of gases, providing a significant tool in environmental monitoring and analysis (Nacapricha et al., 2020).


Analysis Note

Assay (alkalimetric)≥ 25.0 %Color≤ 10 HazenBromide (Br)≤ 50 ppmFree chlorine (Cl)≤ 0.5 ppmSulfate (SO₄)≤ 1 ppmSulfite (SO₃)≤ 1 ppmAl (Aluminium)≤ 0.050 ppmAs (Arsenic)≤ 0.01 ppmBa (Barium)≤ 0.020 ppmBe (Beryllium)≤ 0.020 ppmBi (Bismuth)≤ 0.100 ppmCa (Calcium)≤ 0.500 ppmCd (Cadmium)≤ 0.010 ppmCo (Cobalt)≤ 0.010 ppmCr (Chromium)≤ 0.020 ppmCu (Copper)≤ 0.020 ppmFe (Iron)≤ 0.200 ppmGe (Germanium)≤ 0.050 ppmK (Potassium)≤ 0.100 ppmLi (Lithium)≤ 0.010 ppmMg (Magnesium)≤ 0.100 ppmMn (Manganese)≤ 0.010 ppmMo (Molybdenum)≤ 0.020 ppmNH₄ (Ammonium)≤ 1 ppmNa (Sodium)≤ 0.500 ppmNi (Nickel)≤ 0.020 ppmPb (Lead)≤ 0.020 ppmSr (Strontium)≤ 0.010 ppmTi (Titanium)≤ 0.100 ppmTl (Thallium)≤ 0.050 ppmV (Vanadium)≤ 0.010 ppmZn (Zinc)≤ 0.050 ppmZr (Zirconium)≤ 0.100 ppmResidue on ignition (as SO₄)≤ 5 ppmMinimum shelf life: see product label

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service