Skip to Content
Merck
All Photos(1)

Key Documents

1.00071

Millipore

CCDA Selective Supplement

for Campylobacter spp., pkg of 10 vials, For the preparation of Campylobacter Blood Free Selective Agar

Synonym(s):

Blood Free Campylobacter Medium Selective Supplement, Campylobacter Blood-Free Medium Selective Supplement, Campylobacter Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

Agency

ISO 10272
ISO

Quality Level

form

lyophilized powder

composition

Amphotericin B, 5 mg
Cefoperazone, 16 mg

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

pH

7.0-7.4 (25 °C, 40 g/L in H2O, suspension)

solubility

40 g/L

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.

General description

CCDA Selective Supplement is a mixture of two antibiotics (Amphotericin B & Cefoperazone) in lyophilized form. Amphotericin B largely reduces the growth of yeasts and molds, while Cefoperazone especially inhibits the growth of Enterobacteriaceae.

Application

CCDA Selective Supplement is an additive used during the preparation of Campylobacter Blood Free Selective Agar for the enrichment of Campylobacter from foods.

Linkage

Replaces: 1.00071

Analysis Note

Typical composition (per vial): Amphotericin B 5.0 mg; Cefoperazone 16.0 mg.

Growth promotion test in accordance with ISO 11133.
Supplement tested with base Art. 1.00070.
Growth
- Campylobacter coli ATCC 43478 (WDCM 00004):
good
- Campylobacter jejuni ATCC 33291 (WDCM 00005): good
- Campylobacter jejuni ATCC 29428 (WDCM 00156): good
- Staphylococcus aureus ATCC 25923 (WDCM 00034): no growth
- Escherichia coli ATCC 25922 (WDCM 00013): inhibited growth
- Escherichia coli ATCC 8739 (WDCM 00012): inhibited growth
Incubation: 44 hrs. 41.5 °C with Anaercult C. E.coli and Staph.aur. aerobic.
Strains tested with three-loop-streak.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service