Skip to Content
Merck
All Photos(4)

Key Documents

07-579

Sigma-Aldrich

Anti-REST Antibody

Upstate®, from rabbit

Synonym(s):

Neural-restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, neuron restrictive silencer factor, repressor binding to the X2 box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... REST(5978)
mouse ... Rest(19712)
rat ... Rest(83618)

General description

RE1-silencing transcription factor (UniProt: Q13127; also known as REST, Neural-restrictive silencer factor, X2 box repressor) is encoded by the REST (also known as NRSF, XBR) gene (Gene ID: 5978) in human. REST is a transcriptional repressor that acts by binding a DNA sequence element called the neuron-restrictive silencer element (NRSE). It is ubiquitously expressed, and higher expression is found in the tissues of the lymphocytic compartment, including spleen, thymus, peripheral blood lymphocytes, and ovary. It is also found in undifferentiated neuronal progenitor cells and is believed to acts as a master negative regular of neurogenesis. Expression of REST is reported to be cell cycle-dependent with reduced levels in the G2 phase. It is involved in maintaining the quiescent state of adult neural stem cells and prevents their premature differentiation into mature neurons. REST restricts the expression of neuronal genes by associating with two distinct corepressors, SIN3A and RCOR1, which in turn recruit histone deacetylase to the promoters of REST-regulated genes. REST also mediates repression by recruiting the BHC complex at RE1/NRSE sites, which acts by deacetylating and demethylating specific sites on histones and acting as a chromatin modifier. Mutations in REST gene can lead to Wilms tumors, a pediatric malignancy of the kidney that occurs due to uncontrolled multiplication of renal stem, stromal, and epithelial cells. (Ref.: Mahamdallie, SS., et al. (2015). Nat. Genet. 47(12); 1471-1474; Huang, Z., et al. (2011). Nat. Cell Biol. 13(2); 142-152).

Specificity

This Rabbit polyclonal antibody detects RE1-silencing transcription factor (REST). It targets an epitope within the C-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to 297 amino acids from the C-terminal region of human RE1-silencing transcription factor (REST).

Application

Anti-REST, Cat. No. 07-579, is a rabbit polyclonal antibody that detects REST and is tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Updated information-Rest is glycosylated. From http://amsdottorato.cib.unibo.it/687/
From the summary:
"Finally, the glycosylation pattern of the REST protein was analysed, moving from the observation that the molecular weight calculated on REST sequence is about 116 kDa but using western blotting this transcription factor appears to have distinct apparent molecular weight (see Table 1.1): this difference could be explained by post-translational modifications of the proteins, like glycosylation. In fact recently, several studies underlined the importance of O-glycosylation in modulating transcriptional silencing, protein phosphorylation, protein degradation by proteasome and protein–protein interactions [Julenius et al., 2005; Zachara and Hart, 2006]. Deglycosilating analysis showed that REST protein in SH-SY5Y and HEK293 cells is Oglycosylated and not N-glycosylated. Moreover, using several combination of deglycosilating enzymes it is possible to hypothesize the presence of Gal-β(1-3)-GalNAc residues on the endogenous REST, while β(1-4)-linked galactose residues may be present on recombinant REST protein expressed in HEK293 cells. However, the O-glycosylation process produces an immense multiplicity of chemical structures and monosaccharides must be sequentially hydrolyzed by a series of exoglycosidase. Further experiments are needed to characterize all the post-translational modification of the transcription factor REST." In particular the authors show that REST migrates at 200kd in SH-SY5Y cells.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.Western Blotting Analysis: A 1:1,000 dilution of this antibody detected REST in HEK293 cell lysate.

Target description

~160 kDa observed; 121.87 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in buffer containing 0.014 M phosphate buffer, pH 7.6, 0.175 M NaCl with 0.07% sodium azide with 30% glycerol.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Human Accelerated Region 1 noncoding RNA is repressed by REST in Huntington's disease.
Johnson R, Richter N, Jauch R, Gaughwin PM, Zuccato C, Cattaneo E, Stanton LW
Physiological Genomics null
Nontelomeric splice variant of telomere repeat-binding factor 2 maintains neuronal traits by sequestering repressor element 1-silencing transcription factor.
Zhang, P; Casaday-Potts, R; Precht, P; Jiang, H; Liu, Y; Pazin, MJ; Mattson, MP
Proceedings of the National Academy of Sciences of the USA null
Regulation of neural macroRNAs by the transcriptional repressor REST.
Johnson, R; Teh, CH; Jia, H; Vanisri, RR; Pandey, T; Lu, ZH; Buckley, NJ; Stanton, LW; Lipovich, L
RNA null
L Ferraris et al.
Nucleic acids research, 39(6), e33-e33 (2010-12-21)
There are a variety of in vivo and in vitro methods to determine the genome-wide specificity of a particular trans-acting factor. However there is an inherent limitation to these candidate approaches. Most biological studies focus on the regulation of particular
David Cheishvili et al.
Human molecular genetics, 20(8), 1585-1594 (2011-01-29)
Deficiency in the IKAP/Elp1 protein leads to the recessive sensory autosomal congenital neuropathy which is called familial dysautonomia (FD). This protein was originally identified as a role player in transcriptional elongation being a subunit of the RNAPII transcriptional Elongator multi-protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service