Skip to Content
Merck
All Photos(1)

Documents

07-428-I

Sigma-Aldrich

Anti-phospho eNOS Antibody (Ser1177)

from rabbit, purified by affinity chromatography

Synonym(s):

Nitric oxide synthase, endothelial, Constitutive NOS, cNOS, EC-NOS, Endothelial NOS, eNOS, NOS type III, NOSIII

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology), porcine (based on 100% sequence homology), sheep (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer1177)

Gene Information

human ... NOS3(4846)

General description

NO, produced by nitric oxide synthases (NOS), is a critical signaling molecule involved in many physiological processes including muscle contraction, vasodilation, neuronal transmission, and immune responses. Some of these effects are mediated through reaction with and activation of guanylate cyclase (GC), resulting in increased cGMP production. With a half-life of seconds, NO readily diffuses through the cytosol and cell membrane, allowing it to act inside or adjacent to the cell in which it was produced. Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) & endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. Cytokines such as interferon-gamma (IFN), tumor necrosis factor (TNF), interleukin-1 and -2, and lipopolysaccarides (LPS) cause an increase in iNOS mRNA, protein, and activity levels. Protein kinase C-stimulating agents exhibit the same effect on iNOS activity. Human iNOS is regulated by calcium/calmodulin (in contrast with mouse NOS2).

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to a region near the C-terminus of human eNOS phosphorylated at Ser1177.

Application

Anti-phospho eNOS Antibody (Ser1177) is a highly specific rabbit polyclonal antibody, that targets eNOS/NOS III & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in untreated and VEGF treated HUVEC cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected phospho eNOS (Ser1177) in 10 µg of VEGF treated HUVEC cell lysate.

Target description

~133 kDa observed

Linkage

Replaces: 07-428

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingqiang Wang et al.
Frontiers in cardiovascular medicine, 8, 747497-747497 (2021-10-29)
Background: Extracorporeal cardiac shock waves (ECSW) have great potential in the treatment of coronary heart disease. Endothelial progenitor cells (EPCs) are a class of pluripotent progenitor cells derived from bone marrow or peripheral blood, which have the capacity to migrate
Hien C Nguyen et al.
The Journal of biological chemistry, 299(3), 102921-102921 (2023-01-22)
Circulating fatty acid-binding protein 3 (FABP3) is an effective biomarker of myocardial injury and peripheral artery disease (PAD). The endothelium, which forms the inner most layer of every blood vessel, is exposed to higher levels of FABP3 in PAD or
Andrea Tryfonos et al.
European journal of applied physiology (2022-10-29)
Endothelial dysfunction is an early and integral event in the development of atherosclerosis and coronary artery disease (CAD). Reduced NO bioavailability, oxidative stress, vasoconstriction, inflammation and senescence are all implicated in endothelial dysfunction. However, there are limited data examining associations
Andrea Tryfonos et al.
European journal of sport science, 23(4), 561-570 (2022-02-24)
Although evidence demonstrates the fundamental role of shear stress in vascular health, predominantly through the release of nitric oxide (NO), the mechanisms by which endothelial cells (EC)s sense and transduce shear are poorly understood. In cultured ECs tyrosine phosphorylation of
Reetu R Singh et al.
Hypertension (Dallas, Tex. : 1979), 77(4), 1299-1310 (2021-03-02)
[Figure: see text].

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service