Skip to Content
Merck
All Photos(1)

Key Documents

07-355

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys23) Antibody

serum, Upstate®

Synonym(s):

H3K23Ac, Histone H3 (acetyl K23)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

Saccharomyces cerevisiae, human, vertebrates

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys23)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.



The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Histone H3 acetylated on lysine 23. Does not recognize unacetylated recombinant histone H3

Immunogen

Ovalbumin-conjugated, synthetic peptide (KQLASAcKAARK-C) corresponding to amino acids 18-27 of yeast histone H3 acetylated on lysine 23 with a C-terminal cysteine added for conjugation purposes

Application

Chromatin Immunoprecipitation:
Representative lot data.
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µL of either normal rabbit serum or 2 µL Anti-acetyl-Histone H3 (Lys23)and the Magna ChIP A Kit (Cat. # 17-610). Successful immunoprecipitation of acetyl-Histone H3 (Lys23) associated DNA fragments were verified by qPCR using Control Primers specific for the human GAPDH promoter region as a positive locus, and MyoD primers as a negative locus. Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Western Blot Analysis:
Representative lot data.
Acid extracts from sodium butyrate treated HeLa cells (Lane 1, Catalog # 17-305) and recombinant Histone H3 (Lane 2, Catalog # 14-494) were probed with Anti-acetyl-Histone H3 (Lys23) (1:100,000 dilution).
Arrow indicates acetyl histone H3 (~17 kDa)
Dot Blot:
Representative lot data.
40 ng and 4ng amounts of histone peptides with various modifications (see table 1) were transferred to PVDF membrane and probed with Anti-Acetyl-Histone H3 (Lys23) antibody (1:2000 dilution). Proteins were visualized using a goat anti-rabbit IgG conjugated to HRP and a chemiluminescence detection system. Image from a 60 second exposure is shown.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-acetyl-Histone H3 (Lys23) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, WB to detect acetyl-Histone H3 (Lys23) also known as H3K23Ac, Histone H3 (acetyl K23).

Quality

routinely evaluated by immunoblot on acid extracts from sodium butyrate treated HeLa cells

Target description

17kDa

Physical form

Antiserum
Rabbit antiserum containing 0.05% sodium azide and 30% glycerol

Storage and Stability

2 years at -20°C

Analysis Note

Control
Acid extracts from sodium butyrate treated HeLa cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleosome competition reveals processive acetylation by the SAGA HAT module.
Ringel, AE; Cieniewicz, AM; Taverna, SD; Wolberger, C
Proceedings of the National Academy of Sciences of the USA null
Kevin S Mayer et al.
Plant physiology, 180(1), 342-355 (2019-02-16)
Histone deacetylases remove acetyl groups from histone proteins and play important roles in many genomic processes. How histone deacetylases perform specialized molecular and biological functions in plants is poorly understood. Here, we identify HIGH EXPRESSION OF OSMOTICALLY RESPONSIVE GENES 15
Non-CG methylation patterns shape the epigenetic landscape in Arabidopsis.
Stroud, H; Do, T; Du, J; Zhong, X; Feng, S; Johnson, L; Patel, DJ; Jacobsen, SE
Nature Structural and Molecular Biology null
Fu Huang et al.
Genes & development, 30(10), 1198-1210 (2016-05-21)
KAT6 histone acetyltransferases (HATs) are highly conserved in eukaryotes and are involved in cell cycle regulation. However, information regarding their roles in regulating cell cycle progression is limited. Here, we report the identification of subunits of the Drosophila Enok complex
Kathryn L Huisinga et al.
Genetics, 202(2), 565-582 (2015-12-19)
Heterochromatin is a common DNA packaging form employed by eukaryotes to constitutively silence transposable elements. Determining which sequences to package as heterochromatin is vital for an organism. Here, we use Drosophila melanogaster to study heterochromatin formation, exploiting position-effect variegation, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service