Skip to Content
Merck
All Photos(2)

Key Documents

07-1781

Sigma-Aldrich

Anti-RAD50 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

RAD50 (S. cerevisiae) homolog, RAD50 homolog, RAD50 homolog (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RAD50(10111)
mouse ... Rad50(19360)

General description

RAD50 (previously known as DNA repair protein RAD50) is homologous to Saccharomyces cerevisiae Rad50 and is a member of the structural maintenance of chromosome (SMC) family and is involved in cell growth and viability. RAD50 is known to encode 2 distinct proteins. The RAD50, XRS2/NBS1 complex is required for activation of ATM which will phosphorylate TERF1, thereby promoting telomerase. Along with MRE11 and NBN, RAD50 also forms the MRE11–RAD50–NBS1 (MRN) complex, which is one of the first factors to be localized to the DNA lesion, where it might initially have a structural role by tethering together, and therefore stabilizing, broken chromosomes. MRN also controls ATR, which is required for SMC1A phosphorylation, resulting in chromatid adhesion and DNA damage response. RAD50 is expressed in all tissue but predominantly in the testis and fibroblasts.

Specificity

This antibody recognizes RAD50.

Immunogen

Synthetic peptide corresponding to RAD50.

Application

Immunocytochemistry:
A 1:500 dilution of this antibody was used to detect RAD50 on HeLa and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-RAD50 Antibody (Rabbit Polyclonal Antibody) validated in ICC, WB to detect RAD50 also known as RAD50 (S. cerevisiae) homolog.

Quality

Western Blot Analysis:
A 1:500 dilution of this antibody was used to detect RAD50 on human spleen lysate.

Target description

~154 kDa

Physical form

Affinity purified
Purified rabbit polyclonal antibody precipitated in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.
To reconstitute the antibody, centrifuge the antibody at a moderate speed (5,000 rpm) for 5 minutes. Carefully remove the ammonium sulfate/PBS buffer solution and discard; 10μL of residual ammonium sulfate solution will not affect the re-suspension of the antibody. Do not let the protein pellet dry, as severe loss of antibody reactivity can occur. Re-suspend the antibody pellet in 100L either standard PBS or TBS (pH 7.3-7.5). DO NOT VORTEX. Mix by gentle stirring with a wide pipet tip or gentle finger-tapping. Let the precipitated antibody rehydrate for 1 hour at 4-25°C prior to use. Small particles of precipitated antibody that fail to re-suspend are normal. Vials are overfilled to compensate for any losses.

Analysis Note

Control
Human spleen lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca Schröder-Heurich et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(3), 1331-1341 (2013-12-18)
Faithful chromosome segregation is required for preserving genomic integrity. Failure of this process may entail chromatin bridges preventing normal cytokinesis. To test whether RAD50, a protein normally involved in DNA double-strand break repair, is involved in abnormal cytokinesis and formation
Juan A Montero et al.
Scientific reports, 6, 35478-35478 (2016-10-19)
DNA damage independent of caspase activation accompanies programmed cell death in different vertebrate embryonic organs. We analyzed the significance of DNA damage during the regression of the interdigital tissue, which sculpts the digits in the embryonic limb. Interdigit remodeling involves
M H Yashavarddhan et al.
Frontiers in pharmacology, 8, 750-750 (2017-11-23)
Drug discovery field has tremendously progressed during last few decades, however, an effective radiation countermeasure agent for the safe administration to the victims of radiation exposure is still unavailable. This multi-model study is aimed at elucidating the mechanistic aspects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service