Skip to Content
Merck
All Photos(1)

Key Documents

06-315

Sigma-Aldrich

Anti-KA2/GRIK5 (Kainate Receptor) Antibody

Upstate®, from rabbit

Synonym(s):

Anti-EAA2, Anti-GRIK2, Anti-GluK5, Anti-KA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, rabbit

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIK5(2901)

Specificity

KA2

Immunogen

21 residue synthetic peptide (KTSPPRPRPGPTGPRELTEHE) corresponding to the c-terminus of rat KA2 with a lysine added to the n-terminus

Application

Anti-KA2/GRIK5 (Kainate Receptor) Antibody is an antibody against KA2/GRIK5 (Kainate Receptor) for use in IC, IH, IP & WB.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on rat brain microsomal preparation

Target description

123kDa

Physical form

Format: Purified
Protein G Chromatography
of 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synaptic expression of the high-affinity kainate receptor subunit KA2 in hippocampal cultures
Roche, K. W. and Huganir, R. L.
Neuroscience, 69, 383-393 (1995)
A Herb et al.
Neuron, 8(4), 775-785 (1992-04-01)
A new ionotropic glutamate receptor subunit termed KA-2, cloned from rat brain cDNA, exhibits high affinity for [3H]kainate (KD approximately 15 nM). KA-2 mRNA is widely expressed in embryonic and adult brain. Homomeric KA-2 expression does not generate agonist-sensitive channels
S Mehta et al.
The Journal of biological chemistry, 276(19), 16092-16099 (2001-03-30)
Recent studies have demonstrated that kainate receptors are associated with members of the SAP90/PSD-95 family (synapse-associated proteins (SAPs)) in neurons and that SAP90 can cluster and modify the electrophysiological properties of GluR6/KA2 kainate receptors when co-expressed in transfected cells. In
Functional cooperation between KA2 and GluR6 subunits is involved in the ischemic brain injury.
Hai-Xia Jiang, Qiu-Hua Guan, Dong-Sheng Pei, Guang-Yi Zhang
Journal of Neuroscience Research null
Sanja Selak et al.
Neuron, 63(3), 357-371 (2009-08-15)
Regulation of surface insertion and internalization of AMPA and NMDA receptors has emerged as a key mechanism for the control of synaptic strength. Regulatory elements for synaptic kainate receptors (KARs) are, however, largely undetermined. We have found that SNAP25 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service