Skip to Content
Merck
All Photos(1)

Key Documents

05-613

Sigma-Aldrich

Anti-β-Catenin Antibody, clone 2H4A7

clone 2H4A7, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H4A7, monoclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ctnnb1(12387)

Specificity

β-Catenin

Immunogen

fusion protein corresponding to residues 1-100 of human β-Catenin

Application

Detect β-Catenin also known as beta catenin with Anti-β-Catenin Antibody, clone 2H4A7 (Mouse Monoclonal Antibody), that has been demonstrated to work in IP, WB & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on RIPA lysates from A431, mouse 3T3/A31 or rat L6 cell lysates

Target description

92kDa

Linkage

Replaces: 04-1002

Physical form

Format: Purified
Protein G Chromatography
Protein G purified IgG in buffer containing 0.1M Tris-Glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide

Storage and Stability

Stable 2 years at 2-8°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josiane Sancéau et al.
The Journal of biological chemistry, 278(38), 36537-36546 (2003-07-09)
Enhanced expression of (pro)matrix metalloproteinase-9 (MMP-9) is associated with human tumor invasion and/or metastasis. COH cells derived from a highly invasive and metastatic Ewing's sarcoma constitutively express proMMP-9. Transfection of a double stranded RNA that targets the MMP-9 mRNA into
Ayako Mimata et al.
Cancer science, 102(5), 942-950 (2011-02-01)
Alterations in the E-cadherin gene are associated with sporadic and hereditary diffuse-type gastric cancer. To determine how the loss of function of E-cadherin affects gastric epithelial cell phenotypes, we generated transgenic mice using the Cre-loxP system in which the E-cadherin
A Bowen et al.
Biochemical and biophysical research communications, 479(4), 793-799 (2016-10-01)
The Wnt signalling pathway in beta-cells has been linked to the development of type 2 diabetes. Investigating the impact of a non-canonical Wnt ligand, Wnt4, on beta-cell function we found that in INS-1 cells, Wnt4 was able to completely block Wnt3a
Nandaraj Taye et al.
Matrix biology : journal of the International Society for Matrix Biology, 120, 24-42 (2023-05-16)
Myogenesis is the process that generates multinucleated contractile myofibers from muscle stem cells during skeletal muscle development and regeneration. Myogenesis is governed by myogenic regulatory transcription factors, including MYOD1. Here, we identified the secreted matricellular protein ADAMTS-like 2 (ADAMTSL2) as
Yu-Jen Chiu et al.
International journal of oncology, 52(5), 1465-1478 (2018-03-24)
Osteosarcoma is the most common primary malignancy of the bone and is characterized by local invasion and distant metastasis. Over the past 20 years, long-term outcomes have reached a plateau even with aggressive therapy. Overexpression of insulin-like growth factor 1 receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service