Skip to Content
Merck
All Photos(1)

Documents

05-584

Sigma-Aldrich

Anti-Tie2/TEK Antibody, clone Ab33

clone Ab33, Upstate®, from mouse

Synonym(s):

CD202b antigen, TEK tyrosine kinase, endothelial, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, venous malformations, multip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ab33, monoclonal

species reactivity

human, rat, pig, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TEK(7010)

General description

TIE2 (tyrosine kinase with Ig and EGF homology domains 2) is expressed almost exclusively in endothelial cells in mice, rats and humans. This receptor possesses a unique extracellular domain containing two immunoglobulin-like loops separated by three epidermal growth factor-like repeats that are connected to three fibronectin type III-like repeats. The ligand for the receptor is Angiopoietin 1. Defects in TIE2 are associated with inherited venous malformations; the TIE2 signaling pathway appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis.

Specificity

Recognizes Tie2/TEK, Mr 145kDa

Immunogen

Fusion protein corresponding to residues 1-745 of human Tie2/TEK with a C-terminal His6-tag purified from infected SF-9 cells. Clone Ab33

Application

Immunoprecipitation: This antibody immunoprecipitated Tie2/TEK, as reported by an independent laboratory.

Immunohistochemistry: Reported to detect Tie2/TEK in frozen, Triton-treated sections.
This antibody is not suitable for paraffin-embedded sections.
This Anti-Tie2/TEK Antibody, clone Ab33 is validated for use in WB, IP, IH for the detection of Tie2/TEK.

Quality

Evaluated by western blot on RIPA lysates from HUVEC cells.

Western Blot Analysis: 0.2-1 μg/mL of this antibody detected Tie2/TEK in RIPA lysates from HUVEC cells.

Target description

145 kDa

Physical form

Format: Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide before the addition of glycerol to 30%. Liquid at -20°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tie2 expression and phosphorylation in angiogenic and quiescent adult tissues.
Wong, A L, et al.
Circulation Research, 81, 567-574 (1997)
Krystyna Teichert-Kuliszewska et al.
Circulation research, 98(2), 209-217 (2005-12-17)
Mutations in the bone morphogenetic protein (BMP) receptor-2 (BMPR2) have been found in patients with idiopathic pulmonary arterial hypertension (IPAH); however, the mechanistic link between loss of BMPR2 signaling and the development of pulmonary arterial hypertension is unclear. We hypothesized
Howard Leong-Poi et al.
Circulation research, 101(3), 295-303 (2007-06-23)
Current methods of gene delivery for therapeutic angiogenesis are invasive, requiring either intraarterial or intramuscular administration. A noninvasive method of gene delivery has been developed using ultrasound-mediated destruction of intravenously administered DNA-bearing carrier microbubbles during their microcirculatory transit. Here we
Autocrine role of angiopoietins during megakaryocytic differentiation.
Saulle, E; Guerriero, R; Petronelli, A; Coppotelli, E; Gabbianelli, M; Morsilli et al.
Testing null
K G Peters et al.
British journal of cancer, 77(1), 51-56 (1998-02-12)
Endothelial receptor tyrosine kinases may play important roles in pathological vascular growth, particularly in tumours. In this study, immunohistochemistry was used to evaluate the expression of a novel endothelial receptor tyrosine kinase, Tie2/Tek, in the endothelium of vascular 'hotspots' in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service