Skip to Content
Merck
All Photos(2)

Documents

05-449-C

Sigma-Aldrich

Anti-Bin1, clone 99D (Ascites Free) Antibody

clone 99D, 1 mg/mL, from mouse

Synonym(s):

Myc box-dependent-interacting protein 1, Amphiphysin II, Amphiphysin-like protein, Box-dependent myc-interacting protein 1, Bridging integrator 1, Bin1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

99D, monoclonal

species reactivity

mouse, chicken, human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

chicken ... Bin1(424228)
human ... BIN1(274)
mouse ... Bin1(30948)

General description

Bin1, also known as Myc box-dependent-interacting protein 1, Amphiphysin II, Amphiphysin-like protein, Box-dependent myc-interacting protein 1, Bridging integrator 1, and encoded by the gene BIN1/AMPHL, is another protein involved in the regulation of synaptic vesicle endocytosis and interestingly also acts as tumor suppressor and inhibits malignant cell transformation. BIN1 is expressed in many tissues but has the highest expression in brain and muscle. BIN1 has isoforms and particular isoforms are expressed in particular tissues. For instance BIN1 isoform 11A is only expressed in the brain and found in axon initial segments and nodes of Ranvier. Mutations in BIN1 are associated with a disordered called centronuclear 2 myopathy. BIN1 is localized to nucleus but some isoforms are associated with cytoplasm.

Immunogen

Linear peptide corresponding to human Bin1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Bin1 in human cerebellum and human skeletal muscle myocytes tissue.
Western Blotting Analysis: A representative lot detected Bin1 in primary chicken embryo fibroblasts (Telfer, J.F., et al. (2005). Cellular Signalling. 17:701-8).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Bin1 in C2C12 cell lysate (Wechsler-Reya, R., et al. (1997). Cancer Res. 57:3258-63).
Western Blotting Analysis: A representative lot detected Bin1 in human and rodent cell lines (Wechsler-Reya, R., et al. (1997). Cancer Res. 57:3258-63).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Bin1 in co-immunoprecipitation studies (Sakamuro, D., et al. (1996). Nat. Genet. 14:69-77).
Immunohistochemistry Analysis: A representative lot detected Bin1 in breast and liver carcinoma tissues (Sakamuro, D., et al. (1996). Nat. Genet. 14:69-77).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Bin1 in C2C12 cell lysate (Wechsler-Reya, R. J., et al. (1998). Mol Cell Biol. 18:566-75).
Western Blotting Analysis: A representative lot detected Bin1 in C2C12 cell lysate (Wechsler-Reya, R. J., et al. (1998). Mol Cell Biol. 18:566-75).
Flow Cytometry Analysis: A representative lot detected Bin1 in C2C12 cell lysate (Wechsler-Reya, R. J., et al. (1998). Mol Cell Biol. 18:566-75).
Immunofluorescence Analysis: A representative lot detected Bin1 in C2C12 cells (Wechsler-Reya, R. J., et al. (1998). Mol Cell Biol. 18:566-75).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Bin1, clone 99D (Ascites Free) Antibody is validated for use in Western Blotting and Immunohistochemistry and Immunoprecipitation and Flow Cytometry and Immunofluorescence for the detection of Bin1.

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Bin1 in 10 µg of human skeletal muscle tissue lysate.

Target description

~60 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eline Lemerle et al.
eLife, 12 (2023-04-21)
Excitation-contraction coupling requires a highly specialized membrane structure, the triad, composed of a plasma membrane invagination, the T-tubule, surrounded by two sarcoplasmic reticulum terminal cisternae. Although the precise mechanisms governing T-tubule biogenesis and triad formation remain largely unknown, studies have
Harriet P Lo et al.
The Journal of cell biology, 220(12) (2021-10-12)
The cavin proteins are essential for caveola biogenesis and function. Here, we identify a role for the muscle-specific component, Cavin4, in skeletal muscle T-tubule development by analyzing two vertebrate systems, mouse and zebrafish. In both models, Cavin4 localized to T-tubules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service