Skip to Content
Merck
All Photos(1)

Key Documents

04-1572-I

Sigma-Aldrich

Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8

culture supernatant, clone 3E8, from rat

Synonym(s):

DNA-directed RNA polymerase II subunit RPB1, RNA polymerase II subunit B1, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

3E8, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

ChIP: suitable
ELISA: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer5)

Gene Information

human ... POLR2B(5431)

General description

RPB1 (RNA polymerase II subunit B1) is the catalytic component of RNA polymerase II which synthesizes mRNA and non-coding RNAs. During transcription, elongation is influenced by the phosphorylation status of the C-terminal domain (CTD) of RPB1, which acts as an assembly platform for factors regulating transcription initiation, elongation, termination and mRNA processing. Phosphorylation occurs mainly at residues ′Ser-2′ and ′Ser-5′ of the tandem 7 residue repeats in the C-terminal domain (CTD), which activates Pol II. This phosphorylation also can occur at ′Ser-7′ of the heptapepdtide repeat. This antibody recognizes the ′Ser-5′ CTD residue of RPB1.

Specificity

This antibody recognizes RNA Polymerase II subunit B1 phosphorylated at Ser5 of the C-terminus domain (CTD).

Immunogen

Epitope: Phosphorylated Ser5 at the C-terminus domain (CTD).
Ovalbumin-conjugated linear peptide corresponding to human RNA Polymerase II subunit B1 phosphorylated at Ser5 of the C-terminal domain (CTD).

Application

Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8 is a highly specific rat monoclonal antibody, that targets RNA Polymerase & has been tested in western blotting & ELISA.
Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to immunoprecipitate RNA polymerase II subunit B1 (phospho-CTD Ser-5) in ChIP (Chapman, R., et al. (2007). Science. 318(5857):1780 -1782.).

ELISA Analysis: A representative lot was used by an independent laboratory to detect RNA polymerase II subunit B1 (phospho-CTD Ser-5) in ChIP (Chapman, R., et al. (2007). Science. 318(5857):1780 -1782.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in untreated and lambda phosphotase treated NIH/3T3 cell lysates.

Western Blot Analysis: A 1:2,000 dilution from a representative lot detected RNA polymerase II subunit B1 (phospho-CTD Ser-5) in 10 µg of untreated and lambda phosphotase treated NIH/3T3 cell lysates.

Target description

~220 kDa observed

Linkage

Replaces: 04-1572

Physical form

Rat monoclonal IgG2aκ with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Untreated and lambda phosphotase treated NIH/3T3 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Yu et al.
Nucleic acids research, 47(9), 4462-4475 (2019-03-14)
The general transcription factor P-TEFb, a master regulator of RNA polymerase (Pol) II elongation, phosphorylates the C-terminal domain (CTD) of Pol II and negative elongation factors to release Pol II from promoter-proximal pausing. We show here that P-TEFb surprisingly inhibits
In vivo live imaging of RNA polymerase II transcription factories in primary cells.
Ghamari, Alireza, et al.
Genes & Development, 27, 767-777 (2013)
Xiaohong Zhao et al.
Cell reports, 34(11), 108870-108870 (2021-03-18)
Ibrutinib, a bruton's tyrosine kinase (BTK) inhibitor, provokes robust clinical responses in aggressive mantle cell lymphoma (MCL), yet many patients relapse with lethal Ibrutinib-resistant (IR) disease. Here, using genomic, chemical proteomic, and drug screen profiling, we report that enhancer remodeling-mediated
Jing Gao et al.
Haematologica (2021-06-25)
Despite significant progress in the treatment of patients with diffuse large B-cell lymphoma (DLBCL) and mantle cell lymphoma (MCL), prognosis of patients with relapsed disease remains poor due to the emergence of drug resistance and subsequent disease progression. Identification of
Baishan Jiang et al.
Nature chemical biology, 17(6), 675-683 (2021-03-24)
Cyclin-dependent kinase 12 (CDK12) is an emerging therapeutic target due to its role in regulating transcription of DNA-damage response (DDR) genes. However, development of selective small molecules targeting CDK12 has been challenging due to the high degree of homology between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service