Skip to Content
Merck
All Photos(1)

Key Documents

T-094

Supelco

Tetrahydrocannabivarin (THCV) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O2
Molecular Weight:
286.41
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Shoot®/Snap-N-Spike®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

storage temp.

−70°C

General description

This certified Snap-N-Spike® solution is suitable for THCV testing by HPLC, GC/MS or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and cannabis potency or impurity profiling testing applications. Cerilliant® Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Tetrahydrocannabivarin (THCV) is one of the main cannabinoids that belongs to the class of C21 terpenophenolic compounds. Tetrahydrocannabivarin (THCV) is a cannabinoid found in Cannabis that reportedly has efficacy for the treatment of diabetes and controlling weight gain.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:

  • Multi-analysis of 11 cannabinoids in biomass and extracts of different cannabis varieties by an HPLC-UV method, following the International Conference on Harmonization (ICH) Tripartite Guideline for Validation of Analytical Procedures
  • Extraction of nine cannabinoids from food and beverage samples having hemp seeds, oil, and flour as ingredients followed by their analysis by liquid chromatography-electrospray ionization-QTrap-tandem mass spectrometry LC/ESI‐QTRAP‐MS/MS
  • Simultaneous analysis of five major cannabinoids and their metabolites in human urine and plasma samples by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS)
  • Gas chromatography-mass spectrometry (GC-MS) based analysis of ten main cannabinoids from hemp inflorescences following their silylation and esterification for derivatization
  • Development and validation of an ultra-high performance liquid chromatographic (UHPLC) method coupled to photodiode array and mass spectrometry detectors to determine 11 cannabinoids in various Cannabis Sativa samples
  • Quantitative detection of seven cannabinoids in 70 different oil-based pharmaceutical products by liquid chromatography (LC) coupled to tandem mass spectrometry detector (MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service