Skip to Content
Merck
All Photos(1)

Key Documents

P-081

Supelco

Pyrovalerone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Pyrovalerone hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23NO · HCl
CAS Number:
Molecular Weight:
281.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CCCC(N1CCCC1)C(=O)c2ccc(C)cc2

InChI

1S/C16H23NO.ClH/c1-3-6-15(17-11-4-5-12-17)16(18)14-9-7-13(2)8-10-14;/h7-10,15H,3-6,11-12H2,1-2H3;1H

InChI key

MWRACNBZNVAJHE-UHFFFAOYSA-N

General description

Pyrovalerone, marketed as the stimulant drug Centroton or Thymergix, is used for the clinical treatment of chronic fatigue or lethargy and as an appetite suppressant for weight-loss purposes. The drug has recently appeared in “bath salts” sold in shops across the UK and US.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter C Meltzer et al.
Journal of medicinal chemistry, 49(4), 1420-1432 (2006-02-17)
Dopamine, serotonin, and norepinephrine are essential for neurotransmission in the mammalian system. These three neurotransmitters have been the focus of considerable research because the modulation of their production and their interaction at monoamine receptors has profound effects upon a multitude
C Héron et al.
European journal of pharmacology, 264(3), 391-398 (1994-11-03)
We have studied the ability of various uptake blockers to protect the dopamine neuronal carrier labeled with [3H]GBR 12783 (1-[2-(diphenylmethoxy)ethyl]-4-(3-phenyl-2-(propenyl)-piperazine) against N-ethylmaleimide-induced alkylation, using membrane preparations obtained from rat striatum. Pure uptake inhibitors such as mazindol, pyrovalerone, nomifensine and methylphenidate
H S Shin et al.
Journal of analytical toxicology, 20(7), 568-572 (1996-11-01)
Detection and identification of pyrovalerone and its metabolite, a hydroxylated product, are described. Their identities were confirmed by comparing their mass spectra and gas chromatographic retention times with those of the synthetic standards. The analytical method of pyrovalerone and its
David J Lapinsky et al.
Bioorganic & medicinal chemistry, 17(11), 3770-3774 (2009-05-16)
Non-tropane-based photoaffinity ligands for the dopamine transporter (DAT) are relatively unexplored in contrast to tropane-based compounds such as cocaine. In order to fill this knowledge gap, a ligand was synthesized in which the aromatic ring of pyrovalerone was substituted with
Mark Durham
Emergency medicine journal : EMJ, 28(12), 1059-1060 (2011-03-17)
Since the classification of miaow miaow (mephedrone) as a class B drug in April this year, a new drug is emerging as a so-called 'legal high'. Deaths have already been attributed to ivory wave in different parts of the country.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service