Skip to Content
Merck
All Photos(1)

Key Documents

N-905

Supelco

Nordiazepam solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClN2O
CAS Number:
Molecular Weight:
270.71
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Clc1ccc2NC(=O)CN=C(c3ccccc3)c2c1

InChI

1S/C15H11ClN2O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9H2,(H,18,19)

InChI key

AKPLHCDWDRPJGD-UHFFFAOYSA-N

General description

Nordiazepam is a benzodiazepine prescribed for treatment of anxiety and panic attacks. This analytical reference standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing applications. Nordiazepam is also a primary urinary metabolite of benzodiazepines diazepam and chlrodiazepoxide.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Millefiori et al.
Journal of molecular modeling, 17(2), 281-287 (2010-05-07)
Vertical ionization energies (VIEs) of medazepam, nordazepam and their molecular subunits have been calculated using the electron propagator method in the P3/CEP-31G* approximation. Vertical electron affinities (VEAs) have been obtained with a ∆SCF procedure at the DFT-B3LYP/6-31+G* level of theory.
Laura Mercolini et al.
Talanta, 80(1), 279-285 (2009-09-29)
Diazepam is frequently used as an adjuvant during antidepressant therapy. Recently, some studies have suggested that the treatment with benzodiazepines could have different efficacy in depressed patients as opposed to non-depressed ones. To clarify the matter, a study is currently
Joris C Verster et al.
Sleep medicine reviews, 17(2), 153-159 (2012-08-14)
The use of benzodiazepine receptor agonists can significantly impair driving performance. The aim of this review was to determine if there is a relation between blood concentrations of these drugs and the degree of driving impairment. A literature search was
Shanlin Fu et al.
Journal of analytical toxicology, 34(5), 243-251 (2010-06-10)
beta-Glucuronidase is an enzyme often employed to de-conjugate beta-glucuronides during urinary drug testing for benzodiazepines. It is commonly accepted that use of beta-glucuronidase is a preferred method of hydrolysis over acid-catalyzed hydrolysis, which is known to induce benzodiazepine degradation and
David A Fishbain et al.
Pain medicine (Malden, Mass.), 10(3), 565-572 (2008-11-11)
The objectives of this medicolegal case report are the following: 1) to present details of a chronic pain patient (CPP) who was placed on chronic opioid analgesic therapy (COAT), and subsequently overdosed on multiple drugs, some of which were not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service