Skip to Content
Merck
All Photos(1)

Key Documents

H-090

Supelco

1α,25-Dihydroxyvitamin D2 solution

5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O3
CAS Number:
Molecular Weight:
428.65
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

5 μg/mL in ethanol

application(s)

clinical testing
clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

CC(\C=C\C(C)C(C)(C)O)C1CCC2C(\CCCC12C)=C\C=C3\CC(O)CC(O)C3=C

InChI

1S/C28H44O3/c1-18(9-10-19(2)27(4,5)31)24-13-14-25-21(8-7-15-28(24,25)6)11-12-22-16-23(29)17-26(30)20(22)3/h9-12,18-19,23-26,29-31H,3,7-8,13-17H2,1-2,4-6H3/b10-9+,21-11+,22-12-

InChI key

ZGLHBRQAEXKACO-WQIUIVEWSA-N

General description

Accurate preparation of a quantitative reference is highly critical for accurate assessment of patient Vitamin D levels. This Certified Spiking Solution® is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls and calibrators for LC/UV and LC-MS/MS Vitamin D assays; as system tuning/suitability standards; linearity verification; internal proficiency testing; and for OEM use in preparation of reagents, development of assays and instrument IQ/OQ. The stable, quantitative solution format provides significant benefits to the user including material savings, increased efficiency of labor, and convenience of use. The shelf stable Snap-N-Spike® format allows laboratories to eliminate painstaking weighing operations and provides the ability to spike a known concentration of analyte into their matrix of choice just before use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Henik et al.
Journal of the American Veterinary Medical Association, 215(11), 1644-1649 (2003-10-22)
Rickets was diagnosed in a 1-year-old cat with a history of weakness, osteopenia, and recurrent fractures. Processes causing rickets include vitamin D deficiency caused by inadequate, nutrition, lack of exposure to sunlight, defective metabolism of parent vitamin D to active
Frederick G Strathmann et al.
Clinical chemistry, 57(9), 1279-1285 (2011-07-20)
1α,25-dihydroxy vitamin D [1,25(OH)(2)D] is the active metabolite of vitamin D. Antibody-based detection methods lack specificity, but when combined with isotope dilution/ultra-performance liquid chromatography (UPLC)-tandem mass spectrometry, immunoextraction provides an attractive method for 1,25(OH)(2)D. We developed a method for simultaneous
Richard J Grostern et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 120(5), 607-612 (2002-05-25)
Although calcitriol (1,25-dihydroxycholecalciferol) and vitamin D(2) inhibit retinoblastoma growth in the athymic (nude) mouse xenograft (Y-79 cell line) model of retinoblastoma, they can cause severe toxicity. To examine the toxicity of and dose-dependent response for the inhibition of tumor growth
Shigeaki Kato et al.
Current opinion in nephrology and hypertension, 16(4), 297-304 (2007-06-15)
Vitamin D has diverse biological actions, and consequently the mechanisms behind how it regulates gene transcription are diverse. Unlike its well described positive effects on gene transcription, little is known about how vitamin D induces transcriptional repression. Vitamin D-induced transcriptional
Rachael M Biancuzzo et al.
The Journal of clinical endocrinology and metabolism, 98(3), 973-979 (2013-02-07)
The purpose of this study was to determine 1,25-dihydroxyvitamin D3 [1,25(OH)2D3] and 1,25-dihydroxyvitamin D2 [1,25(OH)2D2] levels in healthy adults consuming 1000 IU vitamin D2 or vitamin D3 per day for 11 weeks. Blood from 34 healthy male and female adults

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service