Skip to Content
Merck
All Photos(1)

Documents

E-076

Supelco

17α-Ethynylestradiol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC=C2C3([H])CCC4(C)C(O)(C#C)CCC4([H])C3([H])CCC2=C1

InChI

1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3

InChI key

BFPYWIDHMRZLRN-UHFFFAOYSA-N

General description

A new Certified Spiking Solution® and certified solution standard suitable for LC/MS and GC/MS applications from clinical and diagnostic testing, endocrinology, and clinical chemistry to environmental testing for EPA Method 1698. 17α-Ethynylestradiol is one of the most commonly used medications and can be found in almost all modern formulations of combined oral contraceptive pills.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 1 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Piérard-Franchimont et al.
Maturitas, 22(2), 151-154 (1995-09-01)
The effects of menopause and hormone replacement therapy (HRT) on the epidermis are largely unknown. The aim of this study was to model and measure the influence of estrogen-transdermal HRT on subtle physiological changes taking place in the epidermis during
Jie Han et al.
Water research, 47(7), 2273-2284 (2013-03-08)
This study demonstrates that ethinylestradiol (EE2), a priority estrogenic contaminant in water, can be rapidly and selectively removed from aqueous solutions using industrial-grade polyamide 612 (PA612) particles as adsorbents. Isothermal studies showed that nonporous low surface area (20 m(2) g(-1)) PA612 particles
Konrad F Koehler et al.
Endocrine reviews, 26(3), 465-478 (2005-04-29)
We have known for many years that estrogen is more than the female hormone. It is essential in the male gonads, and in both sexes, estrogen has functions in the skeleton and central nervous system, on behavior, and in the
Kristina Gemzell-Danielsson et al.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology, 29 Suppl 1, 1-14 (2013-03-06)
There have been numerous attempts to control fertility after unprotected sexual intercourse (UPSI). From very bizarre methods like the vaginal application of Coca Cola to the more serious attempts using calcium antagonists influencing fertility parameters in sperm to hormonal methods
Ahmed M Al-Ansari et al.
Aquatic toxicology (Amsterdam, Netherlands), 132-133, 134-140 (2013-03-19)
We used male goldfish (Carassius auratus) as a model species to determine the uptake, elimination, and bioaccumulation of 17α-ethinylestradiol (EE2). Goldfish were exposed to EE2 via two different routes: water (135ng/L±12.8 standard deviation) for 72h, and food (231ng/g±42 SD) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service