Skip to Content
Merck
All Photos(1)

Key Documents

B-061

Supelco

25B-NBOMe hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22BrNO3 · HCl
Molecular Weight:
416.74
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

BrC1=CC(OC)=C(CCNCC2=C(OC)C=CC=C2)C=C1OC.Cl

InChI

1S/C18H22BrNO3.ClH/c1-21-16-7-5-4-6-14(16)12-20-9-8-13-10-18(23-3)15(19)11-17(13)22-2;/h4-7,10-11,20H,8-9,12H2,1-3H3;1H

InChI key

HUHVUTZPPOFALF-UHFFFAOYSA-N

General description

25B-NBOMe is a second generation 2C amine derivative of the phenethylamine 2C-B. Sold under street names such as New Nexus, N-Bomb, and 2CB this designer drug is a very potent hallucinogen with active doses in the microgram range. Suitable uses for this certified Snap-N-Spike® solution include as starting material for calibrators or controls in 25B-NBOMe testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service