Skip to Content
Merck
All Photos(1)

Documents

A-141

Supelco

Acylcarnitines Mix 1 (C0,C2) hydrochloride solution

10 mg/mL in methanol (each analyte as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO3 · HCl
Molecular Weight:
197.66
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

O-Acetyl-L-carnitine, 10 mg/mL
L-Carnitine hydrochloride, 10 mg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

10 mg/mL in methanol (each analyte as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

multi-component solution

shipped in

wet ice

storage temp.

−70°C

General description

A Certified Spiking Solution® of routinely tested Carnitine and Acetylcarnitine at a concentration of 10 mg/mL. Acylcarnitines are biomarker for many inborn errors of metabolism that cause several metabolic diseases and are routinely tested in newborn screening programs. This multi-component mix is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls, calibrators, and linearity standards for LC-MS/MS assays; as system tuning/suitability standards; linearity verification; and internal proficiency testing. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor. Certified concentration values for each component provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service