Skip to Content
Merck
All Photos(1)

Key Documents

A-046

Supelco

Atropine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Atropine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO3
CAS Number:
Molecular Weight:
289.37
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

N1(C2CCC1CC(C2)OC(=O)C(CO)c3ccccc3)C

InChI

1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3

InChI key

RKUNBYITZUJHSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical toxicology to forensic analysis and pharmaceutical research. Atropine is a pharmaceutical drug used for treatment of cardiac conditions.

For the analysis of the phytotoxins. Tropane alkaloid.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Wen Cheng et al.
International journal of medicinal mushrooms, 15(3), 277-286 (2013-05-15)
In our previous research, Cordyceps militaris (CM) had a hypoglycemic effect in normal rats. In this study we wanted to elucidate whether CM also had an effect on diabetic rats. Twelve rats with streptozotocin-induced diabetes were separated randomly into 2
A I Strelnikova et al.
Bulletin of experimental biology and medicine, 154(2), 180-183 (2013-01-19)
Acetylcholine (5.5×10(-10)-5.5×10(-6)M) accelerated erythrocyte agglutination in men, non-pregnant women in follicular phase of the menstrual cycle, and pregnant women in the first trimester. The effect was blocked with atropine (5.5×10(-6)M). Acetylcholine had no effect on the rate of erythrocyte agglutination
Marija Lukac et al.
European journal of pediatric surgery : official journal of Austrian Association of Pediatric Surgery ... [et al] = Zeitschrift fur Kinderchirurgie, 23(1), 80-84 (2013-02-05)
Evaluation of the effectiveness of oral atropine versus surgical therapy for hypertrophic pyloric stenosis (HPS). A total of 66 consecutive patients with HPS were treated at the University Children's Hospital between January 2006 and December 2011. The diagnosis was initially
Max J Hilz et al.
Stroke, 44(4), 1062-1069 (2013-02-21)
Pharmacological blockade showed sympathetic origin of 0.03 to 0.15 Hz blood pressure (BP) oscillations and parasympathetic origin of 0.15 to 0.5 Hz RR-interval (RRI) oscillations, but has not been used to determine origin of cerebral blood flow velocity (CBFV) oscillations
Yoshizumi Takemura et al.
American journal of physiology. Lung cellular and molecular physiology, 304(6), L428-L437 (2013-01-08)
We and others have shown that epithelial Na(+) channels (ENaC) in alveolar type 2 (AT2) cells are activated by β2 agonists, steroid hormones, elevated oxygen tension, and by dopamine. Although acetylcholine receptors (AChRs) have been previously described in the lung

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service