Skip to Content
Merck
All Photos(2)

Key Documents

855775P

Avanti

18:0 Lyso PC

Avanti Research - A Croda Brand

Synonym(s):

1-octadecanoyl-sn-glycero-3-phosphocholine; PC(18:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H54NO7P
CAS Number:
Molecular Weight:
523.68
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-stearoyl-2-hydroxy-sn-glycero-3-phosphocholine

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 1 g (855775P-1g)
pkg of 1 × 200 mg (855775P-200mg)
pkg of 1 × 25 mg (855775P-25mg)
pkg of 1 × 500 mg (855775P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C26H54NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26(29)32-23-25(28)24-34-35(30,31)33-22-21-27(2,3)4/h25,28H,5-24H2,1-4H3

InChI key

IHNKQIMGVNPMTC-UHFFFAOYSA-N

Application

18:0 Lyso PC has been used:
  • to functionalize carbon nanotubes (CNTs) for the development of methods to attach DNA to CNTs
  • to increase the level of lysophosphatidic acid (LPA) in mouse
  • to induce TNF receptor associated factor 3 interacting protein 2 (TRAF3IP2) expression in endothelial cells (EC)

Packaging

20 mL Clear Glass Screw Cap Vial (855775P-1g)
20 mL Clear Glass Screw Cap Vial (855775P-500mg)
5 mL Amber Glass Screw Cap Vial (855775P-200mg)
5 mL Amber Glass Screw Cap Vial (855775P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

OxLDL induces endothelial dysfunction and death via TRAF3IP2: inhibition by HDL3 and AMPK activators
Valente AJ, et al.
Free radical biology & medicine, 70, 117-128 (2014)
Optimising DNA binding to carbon nanotubes by non-covalent methods
Vanesa Sanz
Carbon, 49(5), 1775-1781 (2011)
Anthony J Valente et al.
Free radical biology & medicine, 70, 117-128 (2014-02-25)
Oxidized low-density lipoprotein (oxLDL) induces endothelial cell death through the activation of NF-κB and AP-1 pathways. TRAF3IP2 is a redox-sensitive cytoplasmic adapter protein and an upstream regulator of IKK/NF-κB and JNK/AP-1. Here we show that oxLDL-induced death in human primary
Otto K Kari et al.
Nanoscale, 12(3), 1728-1741 (2020-01-03)
Methodological constraints have limited our ability to study protein corona formation, slowing nanomedicine development and their successful translation into the clinic. We determined hard and soft corona structural properties along with the corresponding proteomic compositions on liposomes in a label-free
Ieva Ailte et al.
Scientific reports, 6, 30336-30336 (2016-07-28)
Shiga toxin (Stx), an AB5 toxin, binds specifically to the neutral glycosphingolipid Gb3 at the cell surface before being transported into cells. We here demonstrate that addition of conical lysophospholipids (LPLs) with large head groups inhibit Stx binding to cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service