Skip to Content
Merck
All Photos(2)

Key Documents

810189P

Avanti

TopFluor PI(3,4,5)P3

Avanti Research - A Croda Brand 810189P, powder

Synonym(s):

1-oleoyl-2-{12-[4-(dipyrrometheneboron difluoride)butanoyl]amino}dodecanoyl-sn-glycero-3-phosphoinositol-3,4,5-trisphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H108 BF2 N7O23P4
CAS Number:
Molecular Weight:
1420.19
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (with stopper and crimp cap (810189P-50ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810189P

shipped in

dry ice

storage temp.

−20°C

General description

PI(3,4,5)P3 (diacylphosphatidylinositol-3,4,5-trisphosphate) is restricted to the cell membrane of inner leaflet. TopFluor PI(3,4,5)P3 is a fluorescently labeled-phosphoinositide derivative containing saturated acyl chain.
Phosphatidylinositol-3,4,5-trisphosphate (PI(3,4,5)P3) is an inadequate phospholipid present on the inner leaflet of the plasma membrane.

Biochem/physiol Actions

PI(3,4,5)P3 (diacylphosphatidylinositol-3,4,5-trisphosphate) serves as a substrate for PTEN (phosphatase and tensin homolog), a tumor suppressor protein. It has binding affinity towards the active site of phosphatase domain. TopFluor PI(3,4,5)P3 might be useful in monitoring protein induced phosphoinositide clustering. This product is considered photostable than other fluorophores.
Phosphatidylinositol-3,4,5-trisphosphate (PI(3,4,5)P3) plays a key role in inducing the cluster of Syntaxin1A at presynaptic terminals. It also regulates the activity protein kinase B/Akt.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810189P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PI(3,4,5)P3 Engagement Restricts Akt Activity to Cellular Membranes
Ebner M, et al.
Molecular Cell, 65(3), 416-431 (2017)
Synaptic PI(3,4,5)P3 Is Required for Syntaxin1A Clustering and Neurotransmitter Release
Khuong TM, et al.
Neuron, 77(6), 1097-1108 (2013)
Biophysical methods for the characterization of PTEN/lipid bilayer interactions
Harishchandra RK, et al.
Methods, 125-125 (2015)
Biophysical methods for the characterization of PTEN/lipid bilayer interactions
Harishchandra RK, et al.
Methods, 77, 125-135 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service