Skip to Content
Merck
All Photos(1)

Key Documents

810188P

Avanti

TopFluor TMR PI

Avanti Research - A Croda Brand 810188P, powder

Synonym(s):

TopFluor TMR PI, 1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3a,4a-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3-phosphoinositol (Ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (with stopper and crimp cap (810188P-50ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810188P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylinositol (PI) is commonly found in animal tissues and bacteria. PI is soluble in water and chloroform/methanol. TopFluor TMR PI is a tetramethylrhodamine (TMR)-labeled fluorescent phospholipid.

Application

TopFluor TMR PI has been used in fluorescence quenching of 1-palmitoyl-2-{12-[(7-nitro-2−1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphatidylcholine (NBD PC) in ternary mixtures of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphatidylcholine (POPC)/ 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphatidylethanolamine(POPE)/ soy l-α-phosphatidylinositol (PI) vesicles.

Biochem/physiol Actions

TopFluor-labeled lipids have spectral properties similar to tetramethylrhodamine (TMR).

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810188P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipids of membranes: Chemistry, biological role and applications as drug carriers
Hatziantonioy S and Demetzos C
Studies in Natural Products Chemistry, 34, 173-202 (2008)
Yi Wen et al.
Biophysical journal, 114(11), 2630-2639 (2018-06-07)
Phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2 or PIP2), is a key component of the inner leaflet of the plasma membrane in eukaryotic cells. In model membranes, PIP2 has been reported to form clusters, but whether these locally different conditions could give rise to
Reorganization of Ternary Lipid Mixtures of Nonphosphorylated Phosphatidylinositol Interacting with Angiomotin
Kimble-Hill, et al.
The Journal of Physical Chemistry B, 122(35), 8404-8415 (2018)
Ann C Kimble-Hill et al.
The journal of physical chemistry. B, 122(35), 8404-8415 (2018-06-08)
Phosphatidylinositol (PI) lipids are necessary for many cellular signaling pathways of membrane associated proteins, such as angiomotin (Amot). The Amot family regulates cellular polarity, growth, and migration. Given the low concentration of PI lipids in these membranes, it is likely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service