Key Documents
Z758027
BRAND® PLT unit (pipette leak testing unit)
AC/DC input 100 - 240 V AC
Synonym(s):
accuracy of a pipette, calibrating pipettes, pipette accuracy
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
packaging
pkg of 1 ea
manufacturer/tradename
BRAND 703970
Related Categories
General description
Improve process reliability with functional validation of air-displacement pipettes!
The most frequent cause of inaccuracy in piston-operated pipettes is leakage. This arises from damage either to the seals, pistons, or tip cones. Often not detectable by the naked eye, leaks lead to significant volume errors. The BRAND pipette leak tester (PLT unit) for air displacement pipettes finds even the smallest leaks within seconds.
  • Limit values for the commercially available single- and multichannel pipettes in the volume range 1 μL to 10 mL are pre-programmed
  • Testing with and without tips
  • Test results in seconds
  • Patent pending
While the PLT unit cannot replace regular gravimetric testing, daily pipette checks can provide a safeguard during the periods between calibrations. Even the smallest leaks are detected! Process reliability for the pipettes is thus significantly improved.
Unit includes: one adapter each for testing single-channel air-displacement pipettes with tip (mounted) and without tip, 2 plugs (for instrument validation), 3 replacement PE filters for the pipette adapters, universal AC adapter, quality certificate and operating manual.
Features and Benefits
  • Functional testing of your air-displacement pipettes in seconds.
  • Automatic documentation of test results with the PLTconnect software.
  • More security while pipetting.
Other Notes
BR703970 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)