manufacturer/tradename
Aven
total L
4.25 in. (108 mm)
General description
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight, high strength, non-magnetic material.
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
- Lightweight (high strength to weight ratio)
- Zero magnetic retentively
- Temperature resistant to 800 °F
- Chemical and corrosion resistant
- Non-solder adhesive
- Bio-compatible (maintain cell integrity, no inflammatory response)
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)