Z677167
NevoCut™ cut protection gloves
size L
Synonym(s):
safety glove
All Photos(1)
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Recommended Products
material
white (polyamide, glass fibre and HPPE fibre steel)
white gloves
size
L
sterility
non-sterile
feature
latex free
powder-free
reusable
standard type EN388 (cat. II)
packaging
pack of 1 pair
manufacturer/tradename
KCL 927
L × thickness
250 mm × 1.50 mm
General description
Nevocut is the new generation of cut protection gloves combining good sensitivity, flexibility and comfort with the highest level of cut protection (level 5).
The combination of polyamide, glass fiber and high tensile PE fiber guarantees a high cut resistance without lowering the gloves dexterity.
Nevocut is ambidextrous and can be washed several times without loosing its excellent fit.
The combination of polyamide, glass fiber and high tensile PE fiber guarantees a high cut resistance without lowering the gloves dexterity.
Nevocut is ambidextrous and can be washed several times without loosing its excellent fit.
Legal Information
NevoCut is a trademark of KCL GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)