Skip to Content
Merck
All Photos(1)

Key Documents

Z675504

IKA® fixing clips for flasks

AS 2.3 fixing clips, for flask volume 100 ml, 1/cs

Synonym(s):

flask clamps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

feature

for flask size 100 mL

packaging

pack of 1
case of 1 ×

manufacturer/tradename

IKA 1234500

Looking for similar products? Visit Product Comparison Guide

General description

Note: for attaching flasks to HS/KS 260 shaker holding flask attachment.

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

required but not provided


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service