Z567582
Aldrich® Benchclamp™ 2-prong
dual adjustment grippy knobs, size L
Synonym(s):
bench clamps
All Photos(4)
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
Recommended Products
size
L
jaw capacity
0-65 mm
prong W
23 mm
General description
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC