Z528897
Ultrasonic processors 500W and 750W
500W, AC/DC input 110 V AC
Synonym(s):
Ultrasonic homogenizers, Ultrasonic processing equipment
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41101519
NACRES:
NB.74
Recommended Products
manufacturer/tradename
Sonics & Materials VCX500-110V
availability
not available in EU
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The VCX 500 offers 500 watts of power to handle a range of liquid processing applications. This model shares the same microprocessor based operating system and programming screens as the VCX750 system.Using a variety of accessories, the system can process a wide range of sample types and volumes for many different applications.The VCX 500 includes a standard 1/2" diameter probe (50 – 250mL volumes) with replaceable tip.
Applications: Cell disruption in bacteria, spores, and tissues; accelerating catalytic reactions; extractions of serums, toxins, enzymes, and viruses from organic sources; solubilizing difficult compounds; emulsification to 0.01 mm; homogenizing liquids; sample preparation from microscopy and particle size analysis.
- Automatic tuning and frequency control
- Integrated temperature controller from 1°C to 100°C
- Stores up to 10 procedures
- Real time display
- Control process time from 1 second to 10 hours
- Shipped complete with a 1/2 in. (13 mm) probe (630-0220) with replaceable tip, tool kit, and instruction manual
Applications: Cell disruption in bacteria, spores, and tissues; accelerating catalytic reactions; extractions of serums, toxins, enzymes, and viruses from organic sources; solubilizing difficult compounds; emulsification to 0.01 mm; homogenizing liquids; sample preparation from microscopy and particle size analysis.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service