Z263176
Wheaton wide-mouth bottle
capacity 32 oz (1000 mL), white polypropylene cap, amber, vinyl flat liner
Synonym(s):
Wheaton bottle, Wheaton bottles, wide mouth glass
About This Item
Recommended Products
material
round bottle
soda-lime glass bottle
vinyl flat liner
white polypropylene cap
cap size
53-400
Joint
Joints threaded neck (53-400)
sterility
non-sterile
feature
graduations: no
with cap
with cap
packaging
pkg of 12 ea
manufacturer/tradename
WHEATON® W216944
capacity
32 oz (1000 mL)
diam. × H
99 mm × 178 mm
color
amber
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service