Key Documents
Z256668
Kontes® 14/10 MICROFLEX® glassware kits
Standard kit
Synonym(s):
746000-0005
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
manufacturer/tradename
Kimble®
Related Categories
General description
Glassware has 14/10 joints with threaded O-ring cap seals and blue nylon connectors that require minimal clamping and support. Kits contain glassware needed to perform experiments described in the textbooks: Microscale Organic Laboratory and Organic Laboratory Techniques: A Microscale Approach
Standard Kit Components, Qty
A
Air condenser (1)
B 15 mL Centrifuge tube (1)
C Claisen adapter (1)
D Compression cap 20-400 (12)
E Connecting adapter (1)
F Jacketed condenser (1)
G 2 to 3 mL Craig recrystallization tube (1)
H Glass Craig recrystallization plug (1)
I Drying tube (1)
J 5 mL Pear-shaped flask (1)
K 10 mL Round-bottom flask (1)
L Gas delivery tube 20-400 (1)
M GC collection tube (1)
N Hickman distillation head (1)
O 0.1 mL Micro vial (1)
O 3 mL Micro vial (1)
O 5 mL Micro vial (1)
P O-ring, EP, size 112 (8)
Q Septum, PTFE/silicone (4)
R Spin vane (1)
S PE syringe (1)
T Thermometer adapter (1)
U Foam insert set (1)
U Ekono-case (1)
Basic Kit Components, Qty
C
Claisen adapter (1)
D Compression cap 20-400 (7)
F Jacketed condenser (1)
G 2 to 3 mL Craig recrystallization tube (1)
H PTFE Craig recrystallization plug (1)
I Drying tube (1)
K 10 mL Round-bottom flask (1)
N Hickman distillation head (1)
O 3 mL Micro vial (1)
O 5 mL Micro vial (1)
P O-ring, EP, size 112 (5)
Q Septum, PTFE/silicone (2)
T Thermometer adapter, (1)
U Universal inlet/vacuum adapter (1)
Foam insert set (1)
Ekono-case (1)
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
MICROFLEX is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
Kit Components Only
Product No.
Description
  • D. Compression cap, 20-400 12
  • O. 0.1, 3.0, and 5.0 micro vials 1 ea
  • P. O-ring, EP, size 112 8
  • Q. Septum, PTFE/silicone 4
Choose from one of the most recent versions:
Certificates of Analysis (COA)