Key Documents
Z164003
Sigma-Aldrich
Gravity column
Long
About This Item
UNSPSC Code:
23151816
NACRES:
NB.51
Pricing and availability is not currently available.
description
plain top
L × I.D. × O.D.
81 cm × 40 mm × 45 mm
Related Categories
Features and Benefits
  • Provided with a 4mm polytetrafluoroethylene (PTFE) stopcock
  • Has no frit and uses a glass-wool plug and sand
  • Use along with coarse silica gel (e.g., Cat. No. 60744) for fast and general-purpose purification.
Choose from one of the most recent versions:
Certificates of Analysis (COA)