Key Documents
Z147303
Sigma-Aldrich
Lecture bottle adapter, CGA to NPT
CGA 180 male inlet and outlet for 1/4 in. NPT female, brass
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Pricing and availability is not currently available.
material
brass
fitting
CGA 180 male inlet and outlet for 1/4 in. NPT female
Other Notes
The adapter connects a lecture-bottle regulator with a CGA 180 female inlet to a needle valve with 1/4 inch NPT male outlet thread.
Choose from one of the most recent versions:
Certificates of Analysis (COA)