Skip to Content
Merck
All Photos(1)

Key Documents

Z122726

Supelco

Sigma-Aldrich® TLC Plates

silica gel 60 matrix, L × W 20 cm × 20 cm, fluorescent indicator

Synonym(s):

Sigma-Aldrich thin-layer chromatography plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110504
NACRES:
NC.07
Pricing and availability is not currently available.

material

glass support
silica gel 60 matrix

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 25 plates

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

20 cm × 20 cm

layer thickness

250 μm

plate L × W

20 cm × 20 cm

particle size

17 μm

pore size

60 Å medium pore diameter

Looking for similar products? Visit Product Comparison Guide

General description

Thin-layer chromatography (TLC) plates for adsorption, partition, and ion-exchange techniques. Six media choices with glass, polyester, and aluminum backings. Silica-based adsorbent layers in Sigma-Aldrich TLC plates incorporate a polymeric binder; highly purified silica gel plates incorporate a gypsum/polymer binder.
  • Silica gel - for weakly to strongly polar separations, pore size 60 Å
  • High purity silica gel - acid washed for separation of aflatoxins
  • Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose)
  • Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color.
  • Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60Å

Application

Sigma-Aldrich® TLC Plates was used for TLC analysis during GTPase activity assay[1] and lipid extraction/analysis.[2]

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuang-Rung Chang et al.
The Journal of biological chemistry, 282(30), 21583-21587 (2007-06-08)
Mitochondria in cells comprise a tubulovesicular reticulum shaped by dynamic fission and fusion events. The multimeric dynamin-like GTPase Drp1 is a critical protein mediating mitochondrial division. It harbors multiple motifs including GTP-binding, middle, and GTPase effector (GED) domains that are
Heiko Runz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(5), 1679-1689 (2002-03-07)
Generation of amyloid-beta (Abeta) from the amyloid precursor protein (APP) requires proteolytic cleavage by two proteases, beta- and gamma-secretase. Several lines of evidence suggest a role for cholesterol on secretase activities, although the responsible cellular mechanisms remain unclear. Here we

Questions

  1. What compound is used for the fluorescence indicator?

    1 answer
    1. The indicator used for this product is manganese activated zinc silicate with green fluorescence for short-wave UV(254nm).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service