Skip to Content
Merck
All Photos(1)

Key Documents

Z113085

Sigma-Aldrich

Aldrich® glass reference electrode

General purpose,

reference Ag/AgCl

Synonym(s):

Glass Electrode, Reference Electrode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.79

feature

ceramic wick

parameter

flow rate (moderate)
−5-100 °C temperature

Looking for similar products? Visit Product Comparison Guide

General description

The reference electrode is used as a pair with an indication electrode to make a measurement. All types supplied with a 30 in. lead. All reference half-cells have a universal reference pin connector.

Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance

Application

pH, titrations

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service