WHA77001101
Whatman® UNIFILTER® 384 well filter plates
clear polystyrene, non-sterile, dye-terminator, 50 ea
Synonym(s):
384 well plates, Z746363, micro plates, well plates
About This Item
Recommended Products
Product Name
Whatman® UNIFILTER® plates 384, size 384 wells, Dye removal plate, well maximum volume 100 μL, GF/C, clear polystyrene, filter bottom with long drip director
material
conical bottom natural wells
natural polypropylene
polystyrene membrane
feature
GF/C, clear polystyrene, filter bottom with long drip director
packaging
pkg of 50 ea
manufacturer/tradename
Whatman 7700-1101
Whatman Article No. 28420789 (US reference)
size
384 wells
well maximum volume
100 μL
well working volume
100 μL
pore size
0.45 μm
binding type
non-treated surface
Related Categories
General description
Polystyrene microplate for clean-up and cell capture
A high-throughput microplate can facilitate cell capture or dye removal and other sample clean-up processes requiring high recovery. Whatman UNIFILTER microplates are a good choice because of their long drip director and their ability to support gel filtration media.
Features and Benefits
- Usable with gel filtration media, allowing high-throughput removal of dye blobs
- Long drip director prevents well-to-well cross contamination
- Rigid, clear polystyrene microplate can withstand centrifugation
- Membrane media option of DNA binding polymer or glass microfiber
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)