Skip to Content
Merck
All Photos(1)

Key Documents

WHA67047500

Whatman® carbon/HEPA filter capsule

Carbon Cap 75

Synonym(s):

Whatman filters, Z743747, carbon filters, filter carbon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

material

glass fiber membrane (With activated charcol bed)
polypropylene housing

sterility

non-sterile

feature

Carbon Cap 75

packaging

pkg of 1 ea

manufacturer/tradename

Whatman 6704-7500
Whatman Article No. 28420132 (US reference)

diam.

75 mm

pore size

>0.3 μm particle size

fitting

male inlet
male outlet (Sliptip)

Looking for similar products? Visit Product Comparison Guide

General description

Carbon Cap capsule filters are filled with activated carbon and include a pleated glass fiber filter. They are suitable for adsorption of organic vapors from air, and for removing color, organics, and chlorine from water.

Carbon cap for organic and chlorine filtration

Large volume chlorine and organic filtration are facilitated by activated carbon, which adsorbs these molecules. Carbon Cap is a filter filled with high-purity, high-efficiency, acid-washed, activated carbon and a High Efficiency Particulate Air (HEPA) filter. It is made specifically for adsorption of organic vapors, chlorine, and decolorizing liquids.

Application

  • Water, chemical, and reagent purification
  • Removes noxious odors, oil mists, and contaminants
  • Compressed air lines and vacuum pumpsInstrument outlet exhausts
  • Eliminates a potential health hazard from the workplace

Features and Benefits

  • Activated carbon acts as an adsorption media for color, organics, and chlorine
  • Pleated glass microfiber filter structure increases surface area, resulting in high efficiency
  • Retains 99.97% of particles greater than 0.3 µm

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service