Skip to Content
Merck
All Photos(1)

Key Documents

W530098

Sigma-Aldrich

Naringenin

natural (US), 98%

Synonym(s):

(±)-Naringenin, (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4′,5,7-Trihydroxyflavanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
FEMA Number:
4797
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
natural (US)
biological source:
Citrus paradisi Macf.
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

Citrus paradisi Macf.

grade

natural (US)

Assay

98%

mp

247-250 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

SMILES string

OC1=CC=C(C=C1)C(C2)OC3=CC(O)=CC(O)=C3C2=O

InChI

1S/C15H12O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-6,13,16-18H,7H2

InChI key

FTVWIRXFELQLPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naringenin is a bioactive flavonoid found in citrus and grapefruit.[1]

Application

Naringenin has been used in a study to compare its antigenotoxic and antioxidative activity with that of naringenin-oxime.[1]

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xingyuan Shi et al.
Journal of cellular and molecular medicine, 25(5), 2563-2571 (2021-02-02)
Lung cancer is one of the major cause for high-death rate all over the world, due to increased metastasize and difficulties in diagnosis. Naringenin is naturally occurring flavonoid found in various fruits including tomatoes, citrus fruit and figs. Naringenin is
Antigenotoxic and antioxidant potentials of newly derivatized compound naringenin-oxime relative to naringenin on human mononuclear cells
Kocyigit A, et al
Drug and Chemical Toxicology, 39(1), 66-73 (2016)
Ruibing Chen et al.
Frontiers in plant science, 9, 789-789 (2018-07-18)
Scutellarin, a flavonoid 7-O-glucuronide, is an essential bioactive compound of Erigeron breviscapus (Vaniot) Hand.-Mazz. used for the treatment of cerebrovascular diseases. However, due to overexploitation and overuse, E. breviscapus is facing the problems of extinction and habitat degradation. In this
Nada Oršolić et al.
European journal of nutrition, 53(5), 1217-1227 (2013-11-26)
Reactive oxygen species play a role in a number of degenerative conditions including osteoporosis. Flavonoids as phyto-oestrogens exert physiological effects against oxidative stress diseases. We developed a retinoic acid-induced bone loss model of rats to assess whether flavonoids and alendronate
Senem Kamiloglu et al.
Journal of the science of food and agriculture, 94(11), 2225-2233 (2014-01-01)
In order to investigate the effect of home processing on the bioaccessibility of health-related constituents of tomatoes, total lycopene, phenolics, flavonoids and antioxidant capacity were determined from seven different tomato products using an in vitro gastrointestinal digestion model. Additionally, the

Questions

  1. Hi I was looking at your website to order the Naringenin (W530098), but the you have option that 54.6$, how much is the quantity of this ample? It is not mentioned. Best regards, Reza

    1 answer
    1. The SAMPLE-K quantities vary from product to product and lot to lot. There is no established quantity assigned. These are typically gram quantities intended for use in formulation evaluation, however, some more expensive compounds may contain smaller amounts.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service