All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H12O3S
CAS Number:
Molecular Weight:
200.25
FEMA Number:
4538
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.143
NACRES:
NA.21
Organoleptic:
earthy; sulfurous
grade:
FG
Halal
Halal
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Halal
Agency
meets purity specifications of JECFA
reg. compliance
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
Assay
≥98%
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
earthy; sulfurous
SMILES string
S(CCC(=O)OC)Cc1[o]ccc1
InChI
1S/C9H12O3S/c1-11-9(10)4-6-13-7-8-3-2-5-12-8/h2-3,5H,4,6-7H2,1H3
InChI key
MXXNUXUTQGTBGJ-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service