Skip to Content
Merck
All Photos(1)

Key Documents

W382501

Sigma-Aldrich

Diethyl sulfide

98%, FG

Synonym(s):

1,1′-Thiobisethane, Ethyl sulfide, NSC 75157

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2S
CAS Number:
Molecular Weight:
90.19
FEMA Number:
3825
Beilstein:
1696909
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.113
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

vapor pressure

105 mmHg ( 37.7 °C)

Assay

98%

refractive index

n20/D 1.442 (lit.)

bp

90-92 °C (lit.)

mp

−100 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

SMILES string

CCSCC

InChI

1S/C4H10S/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

LJSQFQKUNVCTIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beer Singh et al.
Journal of hazardous materials, 139(1), 38-43 (2006-08-26)
Breakthrough behavior of diethyl sulphide vapors on carbon systems such as active carbon, NaOH/CrO3/C, NaOH/CrO3/EDA/C and RuCl3/C has been studied by using modified Wheeler equation and the same was used to calculate the pseudo-first-order rate constant (kv) and kinetic saturation
R Singh et al.
Chemical research in toxicology, 10(1), 70-77 (1997-01-01)
Postlabeling can be one of the most sensitive methods for the measurement of DNA adducts. However, for the determination of alkylated adducts, essential requirements are standards which must be fully chemically characterized. In order to develop a postlabeling assay for
Taylor C Hayward et al.
Journal of chromatography. A, 1105(1-2), 66-70 (2005-09-30)
A recently reported micro-flame photometric detector (microFPD) has been examined in greater detail for its sulfur response characteristics. While supporting an "upside down" flame on a stainless steel capillary burner (delivering oxygen) in a counter flowing stream of premixed hydrogen
V Vodyanoy et al.
Neuroscience letters, 73(3), 253-258 (1987-01-27)
The steady-state conductance of planar bimolecular lipid membranes (BLMs) modified with rat olfactory epithelial homogenate (ROH) becomes sensitive to very low concentrations of odorant in the presence of adenosine triphosphate (ATP) and guanosine triphosphate (GTP). The chemosensitivity is not observed
Andrea Belancic Majcenovic et al.
Journal of agricultural and food chemistry, 50(23), 6653-6658 (2002-10-31)
Ethanethiol and diethyl disulfide (DEDS) most often occurred at levels above their olfactive threshold in wines with nauseous sulfur-linked smells. As ethanethiol is very oxidizable and chemically reactive, a stable isotopic dilution analysis of both ethanethiol and its disulfide in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service