Skip to Content
Merck
All Photos(1)

Key Documents

W331503

Sigma-Aldrich

2-Nonanol

≥97%

Synonym(s):

Heptyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH(OH)CH3
CAS Number:
Molecular Weight:
144.25
FEMA Number:
3315
Beilstein:
1719466
EC Number:
Council of Europe no.:
4149
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.087
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Assay

≥97%

refractive index

n20/D 1.431 (lit.)

bp

193-194 °C (lit.)

mp

−36-−35 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; green; waxy; citrus

SMILES string

CCCCCCCC(C)O

InChI

1S/C9H20O/c1-3-4-5-6-7-8-9(2)10/h9-10H,3-8H2,1-2H3

InChI key

NGDNVOAEIVQRFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Nonanolis is a simple alcohol with a fruity, green, fatty odor . It is used in the food and perfume industries as flavoring agents and fragrances .

Application

2-Nonanol is a flavor component that contributes to the overall aroma and flavor profile of blue cheese .

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitation, evaluation, and effect of certain microorganisms on flavor components of blue cheese
Anderson DF, et al
Journal of Agricultural and Food Chemistry, 14(3), 241-245 (1966)
Catalytic deoxygenation of bio-based 3-hydroxydecanoic acid to secondary alcohols and alkanes
Mensah JB, et al.
Green Chemistry, 22(11), 3522-3531 (202)
Analysis of free and bound aroma compounds of pomegranate (Punica granatum L)
Tripathi J, et al.
LWT--Food Science and Technology, 59(1), 461-466 (2014)
Fabien Démares et al.
Pharmacology, biochemistry, and behavior, 124, 137-144 (2014-06-10)
Glutamate-gated chloride channels (GluCl) belong to the cys-loop ligand-gated ion channel superfamily and their expression had been described in several invertebrate nervous systems. In the honeybee, a unique gene amel_glucl encodes two alternatively spliced subunits, Amel_GluCl A and Amel_GluCl B.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service