Skip to Content
Merck
All Photos(2)

Documents

W316620

Sigma-Aldrich

Nootkatone

≥98%, FG

Synonym(s):

(+)-Nootkatone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O
CAS Number:
Molecular Weight:
218.33
FEMA Number:
3166
Beilstein:
4676969
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
Flavis number:
07.089
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

grapefruit; orange; sweet; woody; citrus

storage temp.

2-8°C

SMILES string

C[C@@H]1CC(=O)C=C2CC[C@H](C[C@@]12C)C(C)=C

InChI

1S/C15H22O/c1-10(2)12-5-6-13-8-14(16)7-11(3)15(13,4)9-12/h8,11-12H,1,5-7,9H2,2-4H3/t11-,12-,15+/m1/s1

InChI key

WTOYNNBCKUYIKC-JMSVASOKSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Regulation of T Lymphocyte Functions through Calcium Signaling Modulation by Nootkatone.: This study explores how nootkatone modulates calcium signaling to regulate T lymphocyte functions, highlighting its potential in immunotherapy (Lee et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Nagoor Meeran et al.
Molecules (Basel, Switzerland), 25(23) (2020-12-04)
In the present study, we assessed whether nootkatone (NKT), a sesquiterpene in edible plants, can provide protection against dyslipidemia, intramyocardial lipid accumulation, and altered lipid metabolism in a rat model of myocardial infarction (MI) induced by subcutaneous injections of isoproterenol
Lena van Giesen et al.
Cell, 183(3), 594-604 (2020-10-31)
Animals display wide-ranging evolutionary adaptations based on their ecological niche. Octopuses explore the seafloor with their flexible arms using a specialized "taste by touch" system to locally sense and respond to prey-derived chemicals and movement. How the peripherally distributed octopus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service