Skip to Content
Merck
All Photos(1)

Key Documents

W308218

Sigma-Aldrich

2-Tridecenal

predominantly trans, ≥95%, stabilized, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH=CHCHO
CAS Number:
Molecular Weight:
196.33
FEMA Number:
3082
EC Number:
Council of Europe no.:
2011
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.078
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥95%

contains

synthetic, α-tocopherol as stabilizer

refractive index

n20/D 1.459 (lit.)

bp

115-118 °C/10 mmHg (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

waxy; citrus

SMILES string

[H]C(=O)\C([H])=C(/[H])CCCCCCCCCC

InChI

1S/C13H24O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h11-13H,2-10H2,1H3/b12-11+

InChI key

VMUNAKQXJLHODT-VAWYXSNFSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service