Skip to Content
Merck
All Photos(1)

Key Documents

W306509

Sigma-Aldrich

Thyme oil

white, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3065
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
Thymus vulgaris and/or Thymus zygis oil
food allergen:
no known allergens

biological source

Thymus vulgaris and/or Thymus zygis oil

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 182.2

optical activity

[α]20/D −1°, neat

origin

France origin

refractive index

n20/D 1.502

bp

195 °C

density

0.917 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

linalool, citral (neral+geranial), limonene (sum of D, L and DL), geraniol, α-pinene and β-pinene

Organoleptic

herbaceous; camphoraceous

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Thyme.

Application


  • Obliteration of H. pylori infection through the development of a novel thyme oil laden nanoporous gastric floating microsponge.: This study presents an innovative application of thyme oil encapsulated within nanoporous microsponges designed to target and eradicate Helicobacter pylori in the stomach, demonstrating the potential of natural compounds in medical applications (Jafar et al., 2024).

  • Experimental trials to assess the immune modulatory influence of thyme and ginseng oil on NDV-vaccinated broiler chickens.: This research evaluates the immunomodulatory effects of thyme oil on vaccinated poultry, suggesting its utility in enhancing immune responses in agricultural practices (Hassanin et al., 2024).

  • Antimicrobial polysaccharide hydrogels embedded with methyl-β-cyclodextrin/thyme oil inclusion complexes for exceptional mechanical performance and chilled chicken breast preservation.: Highlights the development of polysaccharide-based hydrogels incorporating thyme oil for food preservation, showcasing its antibacterial properties and potential for extending the shelf life of perishable goods (Chen et al., 2024).

  • Corn cob nanocellulose packaging for increasing the shelf life of food products.: Investigates the role of thyme oil in biodegradable packaging solutions, enhancing the antimicrobial properties of corn cob nanocellulose composites, which could revolutionize sustainable packaging technologies (Cheran et al., 2024).

  • Evaluation of the effectiveness of natural extract as a substituent for synthetic preservatives and antioxidants in pharmaceutical preparations.: Examines thyme oil′s efficacy as a natural preservative in pharmaceutical products, promoting its application as a safer alternative to synthetic additives (Al-Rimawi et al., 2024).

Biochem/physiol Actions

Taste at 50 ppm

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Xue et al.
International journal of food microbiology, 210, 1-8 (2015-06-18)
Emulsions of essential oils are investigated as potential intervention strategies to improve food safety and are preferably prepared from generally-recognized-as-safe emulsifiers. Stable thyme oil nanoemulsions can be prepared using combinations of sodium caseinate (NaCas) and soy lecithin. The objective of
Jelica Lazarević et al.
Insects, 11(9) (2020-08-28)
The bean weevil Acanthoscelides obtectus Say (Coleoptera: Chrysomelidae: Bruchinae) can cause significant losses in production of its primary host common bean Phaseolus vulgaris L. To avoid bean protection with environmentally risky chemical insecticides and provide sustainable and safe production of
Siqi Li et al.
Food chemistry, 339, 128016-128016 (2020-11-07)
The aim of this study was to develop a thyme oil emulsion with good physicochemical properties and antibacterial activity. Initially, oil-in-water emulsions containing whey protein-coated essential oil droplets were prepared by high-pressure homogenization. The double-layer emulsions were formed around the
Victor Ryu et al.
Food chemistry, 245, 104-111 (2017-12-31)
The objective of this research was to study the impact of ripening inhibitor level and type on the formation, stability, and activity of antimicrobial thyme oil nanoemulsions formed by spontaneous emulsification. Oil-in-water antimicrobial nanoemulsions (10 wt%) were formed by titrating a
L L Dunn et al.
Journal of food protection, 82(1), 159-163 (2019-02-01)
Alternative postharvest sanitizers to chlorine are of increasing interest for many organic growers and consumers. An emulsion of clove bud oil (CBO; 0.2 and 0.5%) or thyme oil (0.2 and 0.5%) was evaluated as a sanitizer for produce washing against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service