Skip to Content
Merck
All Photos(1)

Key Documents

W288608

Sigma-Aldrich

2-Phenylpropionaldehyde

≥95%, FCC, FG

Synonym(s):

2-Phenylpropanal, Hydratropaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H5)CHO
CAS Number:
Molecular Weight:
134.18
FEMA Number:
2886
Beilstein:
1905601
EC Number:
Council of Europe no.:
126
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.038
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.517 (lit.)

bp

92-94 °C/12 mmHg (lit.)

density

1.002 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; green; floral

storage temp.

2-8°C

SMILES string

[H]C(=O)C(C)c1ccccc1

InChI

1S/C9H10O/c1-8(7-10)9-5-3-2-4-6-9/h2-8H,1H3

InChI key

IQVAERDLDAZARL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Phenylpropionaldehyde is used in perfumes for its green hyacinth odor.

Application


  • Cytotoxicity, early safety screening, and antimicrobial potential of minor oxime constituents of essential oils and aromatic extracts.: Explores the safety and effectiveness of 2-Phenylpropionaldehyde among other compounds in essential oils, highlighting its potential antimicrobial properties and implications for food safety and preservation (Strub DJ et al., 2022).

  • Spectroscopic Evidence for a Cobalt-Bound Peroxyhemiacetal Intermediate.: This study provides spectroscopic evidence of a cobalt-bound intermediate in reactions involving 2-Phenylpropionaldehyde, advancing our knowledge of chemical reaction mechanisms and catalysis (Cho J et al., 2021).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

tert-Butyldimethylsilylated cyclodextrins: versatile chiral stationary phases in capillary gas chromatography.
Maas B, et al.
Journal of Chromatographic Science, 33(5), 223-228 (1995)
M Popović et al.
Chemical research in toxicology, 17(12), 1568-1576 (2004-12-21)
Felbamate (2-phenyl-1,3-propanediol dicarbamate, FBM) can cause aplastic anemia and hepatotoxicity. The mechanism of FBM-induced toxicities is unknown; however, it has been proposed that 2-phenylpropenal, a reactive metabolite of FBM, is responsible. The pathway leading to this metabolite involves hydrolysis of
Shane G Roller et al.
Chemical research in toxicology, 15(6), 815-824 (2002-06-18)
Felbamate is an anti-epileptic drug associated with hepatotoxicity and aplastic anemia. These toxicities are believed to be mediated by the formation of the reactive species 2-phenylpropenal. 4-Hydroxy-5-phenyl-[1,3]oxazinan-2-one is a metabolic precursor for 2-phenylpropenal. 4-Hydroxy-5-phenyl-[1,3]oxazinan-2-one exists in equilibrium with 3-oxo-2-phenylpropyl carbamate
C D Thompson et al.
Chemical research in toxicology, 9(8), 1225-1229 (1996-12-01)
We propose that 3-carbamoyl-2-phenylpropionaldehyde is an intermediate in the metabolism of felbamate, an anti-epileptic drug with a unique profile of the therapeutic activity, and undergoes a cascade of chemical reactions responsible for the toxic properties of the parent drug. To
Robert J Parker et al.
Chemical research in toxicology, 18(12), 1842-1848 (2005-12-20)
Evidence has been presented suggesting that a reactive metabolite, 2-phenylpropenal (ATPAL), may be responsible for the toxicities observed during therapy with the antiepileptic drug felbamate (FBM). Formation of ATPAL from its unstable immediate precursor, 3-carbamoyl-2-phenylpropionaldedhyde (CBMA) requires the loss of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service